Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 80210714-80210963 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTOGL
Entrez Gene ID
283310
Gene Name
otogelin like
Gene Aliases
C12orf64, DFNB84B
UniGene
Hs.723594
Gene Chromosome Location
Chr.12: 80130377-80379090 on Build GRCh38
Transcripts
NM_173591.3 XM_005268802.3 XM_011538191.1

SNP Detail

SNP ID Chromosome Location
rs921533147 Chr.12: 80210848-80210848 on Build GRCh38
rs1029905588 Chr.12: 80210884-80210884 on Build GRCh38
rs766428430 Chr.12: 80210871-80210871 on Build GRCh38
rs982929815 Chr.12: 80210895-80210895 on Build GRCh38
rs937996832 Chr.12: 80210940-80210940 on Build GRCh38
rs764093389 Chr.12: 80210882-80210882 on Build GRCh38
rs1020944228 Chr.12: 80210791-80210791 on Build GRCh38
rs190997924 Chr.12: 80210887-80210887 on Build GRCh38
rs753923439 Chr.12: 80210893-80210893 on Build GRCh38
rs762611150 Chr.12: 80210830-80210830 on Build GRCh38
rs895514947 Chr.12: 80210877-80210877 on Build GRCh38
rs147902353 Chr.12: 80210860-80210860 on Build GRCh38
rs186272901 Chr.12: 80210796-80210796 on Build GRCh38
rs1033297877 Chr.12: 80210913-80210913 on Build GRCh38
rs988181669 Chr.12: 80210779-80210779 on Build GRCh38
rs543655587 Chr.12: 80210906-80210906 on Build GRCh38
rs181526687 Chr.12: 80210760-80210760 on Build GRCh38
rs1032810525 Chr.12: 80210755-80210755 on Build GRCh38
rs530891214 Chr.12: 80210930-80210930 on Build GRCh38
rs117411391 Chr.12: 80210867-80210867 on Build GRCh38
rs976995410 Chr.12: 80210824-80210824 on Build GRCh38
rs542712938 Chr.12: 80210840-80210840 on Build GRCh38
rs1009349624 Chr.12: 80210782-80210782 on Build GRCh38
rs779965995 Chr.12: 80210922-80210922 on Build GRCh38
rs958363607 Chr.12: 80210759-80210759 on Build GRCh38
rs896403697 Chr.12: 80210776-80210776 on Build GRCh38
rs752710374 Chr.12: 80210869-80210869 on Build GRCh38
rs760011677 Chr.12: 80210862-80210862 on Build GRCh38
rs903597986 Chr.12: 80210825-80210825 on Build GRCh38
rs949716264 Chr.12: 80210881-80210881 on Build GRCh38
rs565543193 Chr.12: 80210907-80210907 on Build GRCh38
rs932890315 Chr.12: 80210850-80210850 on Build GRCh38
rs766683073 Chr.12: 80210856-80210856 on Build GRCh38

View Full Product Details