Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 38299964-38300207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNJ15
Entrez Gene ID
3772
Gene Name
potassium voltage-gated channel subfamily J member 15
Gene Aliases
IRKK, KIR1.3, KIR4.2
UniGene
Hs.411299
Gene Chromosome Location
Chr.21: 38229915-38303126 on Build GRCh38
Transcripts
NM_001276435.1 NM_001276436.1 NM_001276437.1 NM_001276438.1 NM_001276439.1 NM_002243.4 NM_170736.2 NM_170737.2 XM_005260975.2 XM_006724002.2 XM_011529560.2 XM_011529561.2 XM_017028343.1 XM_017028344.1 XM_017028345.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6207472 c.737C>T p.P246L Chr.21: 38299998-38299998 on Build GRCh38
6449904 c.736C>T p.P246S Chr.21: 38299997-38299997 on Build GRCh38
2845088 c.786C>T p.S262S Chr.21: 38300047-38300047 on Build GRCh38
6449317 c.889C>T p.R297* Chr.21: 38300150-38300150 on Build GRCh38
2845086 c.766C>T p.H256Y Chr.21: 38300027-38300027 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759831724 Chr.21: 38300101-38300101 on Build GRCh38
rs113056416 Chr.21: 38299992-38299992 on Build GRCh38
rs373264618 Chr.21: 38300050-38300050 on Build GRCh38
rs764116801 Chr.21: 38300030-38300030 on Build GRCh38
rs756190563 Chr.21: 38300014-38300014 on Build GRCh38
rs768306794 Chr.21: 38300068-38300068 on Build GRCh38
rs534907364 Chr.21: 38300150-38300150 on Build GRCh38
rs767855624 Chr.21: 38300102-38300102 on Build GRCh38
rs752858227 Chr.21: 38300011-38300011 on Build GRCh38
rs953962349 Chr.21: 38299991-38299991 on Build GRCh38
rs753794382 Chr.21: 38300035-38300035 on Build GRCh38
rs767690385 Chr.21: 38299995-38299995 on Build GRCh38
rs746624220 Chr.21: 38300066-38300066 on Build GRCh38
rs143785199 Chr.21: 38300134-38300134 on Build GRCh38
rs750314102 Chr.21: 38300135-38300135 on Build GRCh38
rs1006346447 Chr.21: 38300039-38300039 on Build GRCh38
rs780437505 Chr.21: 38300082-38300084 on Build GRCh38
rs774909359 Chr.21: 38300091-38300091 on Build GRCh38
rs577377962 Chr.21: 38300044-38300044 on Build GRCh38
rs963923530 Chr.21: 38300183-38300183 on Build GRCh38
rs771410592 Chr.21: 38300081-38300081 on Build GRCh38
rs138958125 Chr.21: 38300080-38300080 on Build GRCh38
rs143222338 Chr.21: 38300178-38300178 on Build GRCh38
rs911145147 Chr.21: 38300180-38300180 on Build GRCh38
rs200619897 Chr.21: 38300119-38300119 on Build GRCh38
rs779581944 Chr.21: 38300053-38300053 on Build GRCh38
rs749926649 Chr.21: 38300043-38300043 on Build GRCh38
rs760835708 Chr.21: 38300114-38300114 on Build GRCh38
rs746781780 Chr.21: 38300174-38300174 on Build GRCh38
rs142343402 Chr.21: 38300116-38300116 on Build GRCh38
rs758161613 Chr.21: 38300138-38300138 on Build GRCh38
rs759576878 Chr.21: 38300062-38300062 on Build GRCh38
rs1028441815 Chr.21: 38299986-38299986 on Build GRCh38
rs376208976 Chr.21: 38300107-38300107 on Build GRCh38
rs762613652 Chr.21: 38300075-38300075 on Build GRCh38

View Full Product Details