Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 4995199-4995424 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPP40
Entrez Gene ID
10799
Gene Name
ribonuclease P/MRP subunit p40
Gene Aliases
RNASEP1, bA428J1.3
UniGene
Hs.511756
Gene Chromosome Location
Chr.6: 4992405-5004063 on Build GRCh38
Transcripts
NM_001286132.1 NM_001286133.1 NM_006638.3 XM_011514253.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5405526 c.968C>T p.S323F Chr.6: 4995202-4995202 on Build GRCh38
1444990 c.940G>A p.V314I Chr.6: 4995230-4995230 on Build GRCh38
6298707 c.960C>A p.S320R Chr.6: 4995210-4995210 on Build GRCh38
3628345 c.938C>T p.S313F Chr.6: 4995232-4995232 on Build GRCh38
1080110 c.908A>G p.E303G Chr.6: 4995262-4995262 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs757238319 Chr.6: 4995301-4995302 on Build GRCh38
rs946065632 Chr.6: 4995368-4995368 on Build GRCh38
rs917107554 Chr.6: 4995251-4995251 on Build GRCh38
rs376437298 Chr.6: 4995238-4995238 on Build GRCh38
rs559922527 Chr.6: 4995273-4995273 on Build GRCh38
rs892030634 Chr.6: 4995329-4995329 on Build GRCh38
rs751695437 Chr.6: 4995279-4995280 on Build GRCh38
rs753159113 Chr.6: 4995231-4995231 on Build GRCh38
rs972572631 Chr.6: 4995243-4995243 on Build GRCh38
rs373844439 Chr.6: 4995301-4995301 on Build GRCh38
rs952846078 Chr.6: 4995330-4995330 on Build GRCh38
rs920377784 Chr.6: 4995325-4995325 on Build GRCh38
rs144676135 Chr.6: 4995271-4995271 on Build GRCh38
rs756472526 Chr.6: 4995234-4995234 on Build GRCh38
rs1010934923 Chr.6: 4995341-4995341 on Build GRCh38
rs75895012 Chr.6: 4995357-4995357 on Build GRCh38
rs779163622 Chr.6: 4995259-4995259 on Build GRCh38
rs373716513 Chr.6: 4995270-4995270 on Build GRCh38
rs757486654 Chr.6: 4995246-4995246 on Build GRCh38
rs12332997 Chr.6: 4995230-4995230 on Build GRCh38
rs371944622 Chr.6: 4995226-4995226 on Build GRCh38
rs536356148 Chr.6: 4995258-4995258 on Build GRCh38
rs776639701 Chr.6: 4995279-4995279 on Build GRCh38
rs917253343 Chr.6: 4995359-4995359 on Build GRCh38
rs762398316 Chr.6: 4995310-4995310 on Build GRCh38
rs1055095275 Chr.6: 4995292-4995292 on Build GRCh38
rs749638819 Chr.6: 4995242-4995242 on Build GRCh38
rs745981946 Chr.6: 4995267-4995267 on Build GRCh38
rs902051731 Chr.6: 4995361-4995361 on Build GRCh38
rs761641531 Chr.6: 4995295-4995295 on Build GRCh38
rs937597785 Chr.6: 4995348-4995348 on Build GRCh38
rs767796077 Chr.6: 4995308-4995309 on Build GRCh38
rs927510504 Chr.6: 4995342-4995342 on Build GRCh38
rs547052558 Chr.6: 4995333-4995333 on Build GRCh38
rs772874277 Chr.6: 4995303-4995303 on Build GRCh38

View Full Product Details