Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 77637020-77637265 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OTP
Entrez Gene ID
23440
Gene Name
orthopedia homeobox
Gene Aliases
-
UniGene
Hs.202247
Gene Chromosome Location
Chr.5: 77628712-77638697 on Build GRCh38
Transcripts
NM_032109.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6396099 c.49G>T p.A17S Chr.5: 77637219-77637219 on Build GRCh38
4655369 c.66C>A p.G22G Chr.5: 77637202-77637202 on Build GRCh38
4650692 c.51C>T p.A17A Chr.5: 77637217-77637217 on Build GRCh38
6513118 c.55G>T p.E19* Chr.5: 77637213-77637213 on Build GRCh38
1438691 c.66C>T p.G22G Chr.5: 77637202-77637202 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780895665 Chr.5: 77637138-77637138 on Build GRCh38
rs759629586 Chr.5: 77637217-77637217 on Build GRCh38
rs760852640 Chr.5: 77637190-77637190 on Build GRCh38
rs370680168 Chr.5: 77637049-77637049 on Build GRCh38
rs374616762 Chr.5: 77637171-77637171 on Build GRCh38
rs767534095 Chr.5: 77637183-77637183 on Build GRCh38
rs757104012 Chr.5: 77637136-77637136 on Build GRCh38
rs868128425 Chr.5: 77637083-77637083 on Build GRCh38
rs764009873 Chr.5: 77637242-77637242 on Build GRCh38
rs141172285 Chr.5: 77637058-77637058 on Build GRCh38
rs1015596217 Chr.5: 77637073-77637073 on Build GRCh38
rs776036160 Chr.5: 77637059-77637059 on Build GRCh38
rs1053321050 Chr.5: 77637130-77637130 on Build GRCh38
rs766407119 Chr.5: 77637107-77637107 on Build GRCh38
rs769218113 Chr.5: 77637164-77637164 on Build GRCh38
rs972368192 Chr.5: 77637220-77637220 on Build GRCh38
rs185198254 Chr.5: 77637056-77637056 on Build GRCh38
rs776648406 Chr.5: 77637216-77637216 on Build GRCh38
rs758279169 Chr.5: 77637115-77637115 on Build GRCh38
rs35484706 Chr.5: 77637229-77637229 on Build GRCh38
rs764026495 Chr.5: 77637128-77637128 on Build GRCh38
rs766641310 Chr.5: 77637200-77637200 on Build GRCh38
rs775012563 Chr.5: 77637170-77637170 on Build GRCh38
rs773450056 Chr.5: 77637172-77637172 on Build GRCh38
rs748839801 Chr.5: 77637159-77637159 on Build GRCh38
rs762272040 Chr.5: 77637068-77637068 on Build GRCh38
rs866920490 Chr.5: 77637088-77637088 on Build GRCh38
rs751301892 Chr.5: 77637131-77637131 on Build GRCh38
rs777590690 Chr.5: 77637047-77637047 on Build GRCh38
rs780725330 Chr.5: 77637050-77637050 on Build GRCh38
rs199908221 Chr.5: 77637038-77637038 on Build GRCh38
rs750666826 Chr.5: 77637090-77637090 on Build GRCh38
rs752604920 Chr.5: 77637108-77637108 on Build GRCh38
rs769309786 Chr.5: 77637065-77637065 on Build GRCh38
rs536092036 Chr.5: 77637078-77637078 on Build GRCh38
rs760802561 Chr.5: 77637091-77637091 on Build GRCh38
rs755630439 Chr.5: 77637153-77637153 on Build GRCh38
rs779383251 Chr.5: 77637155-77637155 on Build GRCh38
rs892010553 Chr.5: 77637134-77637134 on Build GRCh38
rs150715110 Chr.5: 77637067-77637067 on Build GRCh38
rs995976957 Chr.5: 77637154-77637154 on Build GRCh38
rs553012107 Chr.5: 77637145-77637145 on Build GRCh38
rs759234533 Chr.5: 77637052-77637052 on Build GRCh38

View Full Product Details