Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 50061500-50061767 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MLC1
Entrez Gene ID
23209
Gene Name
megalencephalic leukoencephalopathy with subcortical cysts 1
Gene Aliases
LVM, MLC, VL
UniGene
Hs.517729
Gene Chromosome Location
Chr.22: 50059391-50085929 on Build GRCh38
Transcripts
NM_015166.3 NM_139202.2 XM_011530678.1 XM_017028671.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5715720 c.1093G>A p.E365K Chr.22: 50061624-50061624 on Build GRCh38
3842909 c.1113C>T p.V371V Chr.22: 50061604-50061604 on Build GRCh38
6217565 c.1060-4G>T p.? Chr.22: 50061661-50061661 on Build GRCh38
6583172 c.1101C>A p.A367A Chr.22: 50061616-50061616 on Build GRCh38
3555573 c.1103G>A p.W368* Chr.22: 50061614-50061614 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760778847 Chr.22: 50061607-50061607 on Build GRCh38
rs368422170 Chr.22: 50061708-50061708 on Build GRCh38
rs763616187 Chr.22: 50061581-50061581 on Build GRCh38
rs750204522 Chr.22: 50061625-50061625 on Build GRCh38
rs759675682 Chr.22: 50061601-50061601 on Build GRCh38
rs764296487 Chr.22: 50061704-50061704 on Build GRCh38
rs779971307 Chr.22: 50061630-50061630 on Build GRCh38
rs749679723 Chr.22: 50061665-50061665 on Build GRCh38
rs758309611 Chr.22: 50061552-50061552 on Build GRCh38
rs762746384 Chr.22: 50061705-50061705 on Build GRCh38
rs200273593 Chr.22: 50061606-50061606 on Build GRCh38
rs376679534 Chr.22: 50061707-50061707 on Build GRCh38
rs1054216006 Chr.22: 50061721-50061721 on Build GRCh38
rs771562961 Chr.22: 50061587-50061587 on Build GRCh38
rs755224475 Chr.22: 50061534-50061534 on Build GRCh38
rs41283496 Chr.22: 50061528-50061528 on Build GRCh38
rs111797969 Chr.22: 50061565-50061565 on Build GRCh38
rs758396297 Chr.22: 50061627-50061627 on Build GRCh38
rs972858942 Chr.22: 50061622-50061622 on Build GRCh38
rs765064183 Chr.22: 50061618-50061618 on Build GRCh38
rs142027672 Chr.22: 50061571-50061571 on Build GRCh38
rs185151842 Chr.22: 50061536-50061536 on Build GRCh38
rs985607951 Chr.22: 50061605-50061605 on Build GRCh38
rs936061235 Chr.22: 50061638-50061638 on Build GRCh38
rs137919 Chr.22: 50061535-50061535 on Build GRCh38
rs751390945 Chr.22: 50061631-50061631 on Build GRCh38
rs200459777 Chr.22: 50061603-50061603 on Build GRCh38
rs769559923 Chr.22: 50061649-50061649 on Build GRCh38
rs778640243 Chr.22: 50061542-50061542 on Build GRCh38
rs569220214 Chr.22: 50061666-50061666 on Build GRCh38
rs774935114 Chr.22: 50061598-50061598 on Build GRCh38
rs2072873 Chr.22: 50061538-50061538 on Build GRCh38
rs577593915 Chr.22: 50061544-50061544 on Build GRCh38
rs775050045 Chr.22: 50061695-50061695 on Build GRCh38
rs369175737 Chr.22: 50061572-50061572 on Build GRCh38
rs759921284 Chr.22: 50061678-50061678 on Build GRCh38
rs892995236 Chr.22: 50061741-50061741 on Build GRCh38
rs772235176 Chr.22: 50061688-50061688 on Build GRCh38
rs1050951853 Chr.22: 50061729-50061729 on Build GRCh38
rs780525099 Chr.22: 50061641-50061641 on Build GRCh38
rs761014007 Chr.22: 50061701-50061701 on Build GRCh38
rs531780319 Chr.22: 50061664-50061664 on Build GRCh38
rs1045583103 Chr.22: 50061527-50061527 on Build GRCh38
rs766109625 Chr.22: 50061706-50061706 on Build GRCh38
rs370011599 Chr.22: 50061554-50061554 on Build GRCh38
rs770221748 Chr.22: 50061573-50061573 on Build GRCh38
rs1039686428 Chr.22: 50061689-50061689 on Build GRCh38
rs542817231 Chr.22: 50061574-50061574 on Build GRCh38
rs747871225 Chr.22: 50061642-50061642 on Build GRCh38
rs774946150 Chr.22: 50061670-50061670 on Build GRCh38
rs950188408 Chr.22: 50061712-50061712 on Build GRCh38
rs769367827 Chr.22: 50061577-50061578 on Build GRCh38
rs886975363 Chr.22: 50061731-50061731 on Build GRCh38
rs775411376 Chr.22: 50061696-50061696 on Build GRCh38
rs373419167 Chr.22: 50061636-50061636 on Build GRCh38
rs78563213 Chr.22: 50061564-50061564 on Build GRCh38

View Full Product Details