Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 168399499-168399724 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WWC1
Entrez Gene ID
23286
Gene Name
WW and C2 domain containing 1
Gene Aliases
HBEBP3, HBEBP36, KIBRA, MEMRYQTL, PPP1R168
UniGene
Hs.484047
Gene Chromosome Location
Chr.5: 168292060-168472303 on Build GRCh38
Transcripts
NM_001161661.1 NM_001161662.1 NM_015238.2 XM_005265850.1 XM_005265853.2 XM_011534485.1 XM_011534486.1 XM_011534487.1 XM_011534488.1 XM_011534489.1 XM_011534490.1 XM_011534491.1 XM_017009276.1 XM_017009277.1 XM_017009278.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6198350 c.590+8C>A p.? Chr.5: 168399575-168399575 on Build GRCh38
1580123 c.536T>C p.V179A Chr.5: 168399513-168399513 on Build GRCh38
5961991 c.541C>G p.L181V Chr.5: 168399518-168399518 on Build GRCh38
1328918 c.557A>C p.Q186P Chr.5: 168399534-168399534 on Build GRCh38
5447638 c.556C>T p.Q186* Chr.5: 168399533-168399533 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776911038 Chr.5: 168399581-168399581 on Build GRCh38
rs951385627 Chr.5: 168399571-168399571 on Build GRCh38
rs769032644 Chr.5: 168399567-168399567 on Build GRCh38
rs532274359 Chr.5: 168399682-168399682 on Build GRCh38
rs951204721 Chr.5: 168399606-168399606 on Build GRCh38
rs936735352 Chr.5: 168399694-168399694 on Build GRCh38
rs551181968 Chr.5: 168399669-168399669 on Build GRCh38
rs780262543 Chr.5: 168399527-168399527 on Build GRCh38
rs773754938 Chr.5: 168399593-168399593 on Build GRCh38
rs770272010 Chr.5: 168399592-168399592 on Build GRCh38
rs186758104 Chr.5: 168399685-168399685 on Build GRCh38
rs551749641 Chr.5: 168399684-168399684 on Build GRCh38
rs745998151 Chr.5: 168399524-168399524 on Build GRCh38
rs762207229 Chr.5: 168399589-168399589 on Build GRCh38
rs980979835 Chr.5: 168399664-168399664 on Build GRCh38
rs754131020 Chr.5: 168399530-168399531 on Build GRCh38
rs970003922 Chr.5: 168399626-168399626 on Build GRCh38
rs182414835 Chr.5: 168399595-168399595 on Build GRCh38
rs563220290 Chr.5: 168399678-168399678 on Build GRCh38
rs772086660 Chr.5: 168399526-168399526 on Build GRCh38
rs562275436 Chr.5: 168399546-168399546 on Build GRCh38
rs1019527681 Chr.5: 168399659-168399659 on Build GRCh38
rs747522670 Chr.5: 168399671-168399671 on Build GRCh38

View Full Product Details