Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 30637591-30637838 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
USPL1
Entrez Gene ID
10208
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
C13orf22, D13S106E, bA121O19.1
UniGene
Hs.533831
Gene Chromosome Location
Chr.13: 30617397-30660770 on Build GRCh38
Transcripts
NM_001321532.1 NM_001321533.1 NM_001321534.1 NM_005800.4 XM_005266213.3 XM_006719751.3 XM_017020314.1 XM_017020315.1 XM_017020316.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6358256 c.869-10A>G p.? Chr.13: 30637734-30637734 on Build GRCh38
4973351 c.907G>T p.E303* Chr.13: 30637782-30637782 on Build GRCh38
5715567 c.924G>A p.L308L Chr.13: 30637799-30637799 on Build GRCh38
1300129 c.893C>T p.S298L Chr.13: 30637768-30637768 on Build GRCh38
5747475 c.880delA p.K295fs*10 Chr.13: 30637755-30637755 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs746888718 Chr.13: 30637787-30637787 on Build GRCh38
rs368975955 Chr.13: 30637705-30637705 on Build GRCh38
rs568112638 Chr.13: 30637668-30637668 on Build GRCh38
rs781357660 Chr.13: 30637764-30637764 on Build GRCh38
rs202200438 Chr.13: 30637785-30637785 on Build GRCh38
rs959447279 Chr.13: 30637797-30637797 on Build GRCh38
rs759060589 Chr.13: 30637710-30637710 on Build GRCh38
rs764757157 Chr.13: 30637728-30637728 on Build GRCh38
rs757702984 Chr.13: 30637750-30637750 on Build GRCh38
rs1017393050 Chr.13: 30637693-30637693 on Build GRCh38
rs759925961 Chr.13: 30637787-30637788 on Build GRCh38
rs373743599 Chr.13: 30637709-30637709 on Build GRCh38
rs3759511 Chr.13: 30637717-30637717 on Build GRCh38
rs139481673 Chr.13: 30637765-30637765 on Build GRCh38
rs376022257 Chr.13: 30637703-30637703 on Build GRCh38
rs780296311 Chr.13: 30637777-30637777 on Build GRCh38
rs1040115501 Chr.13: 30637699-30637699 on Build GRCh38
rs1007311405 Chr.13: 30637643-30637643 on Build GRCh38
rs147344218 Chr.13: 30637719-30637719 on Build GRCh38
rs749424075 Chr.13: 30637780-30637780 on Build GRCh38
rs760528828 Chr.13: 30637700-30637700 on Build GRCh38
rs773135521 Chr.13: 30637786-30637786 on Build GRCh38
rs1032806904 Chr.13: 30637788-30637788 on Build GRCh38
rs764831758 Chr.13: 30637713-30637713 on Build GRCh38
rs112210719 Chr.13: 30637789-30637789 on Build GRCh38
rs374186272 Chr.13: 30637744-30637744 on Build GRCh38

View Full Product Details