Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 31409159-31409384 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DROSHA
Entrez Gene ID
29102
Gene Name
drosha ribonuclease III
Gene Aliases
ETOHI2, HSA242976, RANSE3L, RN3, RNASE3L, RNASEN
UniGene
Hs.97997
Gene Chromosome Location
Chr.5: 31400494-31532175 on Build GRCh38
Transcripts
NM_001100412.1 NM_013235.4 XM_005248291.3 XM_005248292.3 XM_005248293.3 XM_005248294.3 XM_011514033.2 XM_017009399.1 XM_017009400.1 XM_017009401.1 XM_017009402.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4619853 c.3681G>T p.A1227A Chr.5: 31409319-31409319 on Build GRCh38
6649333 c.3696G>T p.K1232N Chr.5: 31409304-31409304 on Build GRCh38
6438266 c.3675T>G p.I1225M Chr.5: 31409325-31409325 on Build GRCh38
166534 c.3715A>T p.T1239S Chr.5: 31409285-31409285 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs745328317 Chr.5: 31409303-31409303 on Build GRCh38
rs78912548 Chr.5: 31409242-31409242 on Build GRCh38
rs755814935 Chr.5: 31409241-31409241 on Build GRCh38
rs765666859 Chr.5: 31409197-31409197 on Build GRCh38
rs368252147 Chr.5: 31409319-31409319 on Build GRCh38
rs778381047 Chr.5: 31409262-31409262 on Build GRCh38
rs770020188 Chr.5: 31409337-31409337 on Build GRCh38
rs771619776 Chr.5: 31409312-31409312 on Build GRCh38
rs766559640 Chr.5: 31409208-31409208 on Build GRCh38
rs923348676 Chr.5: 31409292-31409292 on Build GRCh38
rs369208371 Chr.5: 31409196-31409196 on Build GRCh38
rs760627877 Chr.5: 31409350-31409350 on Build GRCh38
rs964728565 Chr.5: 31409239-31409239 on Build GRCh38
rs774891808 Chr.5: 31409238-31409238 on Build GRCh38
rs771539167 Chr.5: 31409328-31409328 on Build GRCh38
rs1047217743 Chr.5: 31409296-31409296 on Build GRCh38
rs990437331 Chr.5: 31409179-31409179 on Build GRCh38
rs777052398 Chr.5: 31409338-31409339 on Build GRCh38
rs755098624 Chr.5: 31409221-31409221 on Build GRCh38
rs776966464 Chr.5: 31409318-31409318 on Build GRCh38
rs371286084 Chr.5: 31409198-31409198 on Build GRCh38
rs1040697864 Chr.5: 31409225-31409225 on Build GRCh38
rs773776370 Chr.5: 31409345-31409345 on Build GRCh38
rs756971051 Chr.5: 31409255-31409255 on Build GRCh38
rs752710538 Chr.5: 31409237-31409237 on Build GRCh38
rs767289743 Chr.5: 31409235-31409235 on Build GRCh38
rs1055580508 Chr.5: 31409358-31409358 on Build GRCh38
rs763303398 Chr.5: 31409203-31409203 on Build GRCh38
rs376142741 Chr.5: 31409211-31409211 on Build GRCh38

View Full Product Details