Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 3280283-3280556 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TSPAN9
Entrez Gene ID
10867
Gene Name
tetraspanin 9
Gene Aliases
NET-5, NET5, PP1057
UniGene
Hs.504517
Gene Chromosome Location
Chr.12: 3077355-3286564 on Build GRCh38
Transcripts
NM_001168320.1 NM_006675.4 XM_011520912.2 XM_011520913.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6861494 c.384C>T p.T128T Chr.12: 3280435-3280435 on Build GRCh38
6451946 c.394G>A p.V132M Chr.12: 3280445-3280445 on Build GRCh38
6519875 c.412T>A p.W138R Chr.12: 3280463-3280463 on Build GRCh38
6341097 c.432+3G>T p.? Chr.12: 3280486-3280486 on Build GRCh38
5342080 c.343G>A p.A115T Chr.12: 3280394-3280394 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765619360 Chr.12: 3280367-3280367 on Build GRCh38
rs758474210 Chr.12: 3280519-3280519 on Build GRCh38
rs375588314 Chr.12: 3280340-3280340 on Build GRCh38
rs71577837 Chr.12: 3280536-3280536 on Build GRCh38
rs200214939 Chr.12: 3280375-3280375 on Build GRCh38
rs781252017 Chr.12: 3280388-3280388 on Build GRCh38
rs71577836 Chr.12: 3280521-3280521 on Build GRCh38
rs756338611 Chr.12: 3280498-3280498 on Build GRCh38
rs774753869 Chr.12: 3280532-3280532 on Build GRCh38
rs767582157 Chr.12: 3280491-3280491 on Build GRCh38
rs771141053 Chr.12: 3280531-3280531 on Build GRCh38
rs189755506 Chr.12: 3280524-3280524 on Build GRCh38
rs537275809 Chr.12: 3280354-3280354 on Build GRCh38
rs771652932 Chr.12: 3280434-3280434 on Build GRCh38
rs992483695 Chr.12: 3280368-3280368 on Build GRCh38
rs754125139 Chr.12: 3280500-3280500 on Build GRCh38
rs371116590 Chr.12: 3280357-3280357 on Build GRCh38
rs778866918 Chr.12: 3280505-3280505 on Build GRCh38
rs778494539 Chr.12: 3280337-3280338 on Build GRCh38
rs762676996 Chr.12: 3280444-3280444 on Build GRCh38
rs60904711 Chr.12: 3280378-3280378 on Build GRCh38
rs66501720 Chr.12: 3280435-3280435 on Build GRCh38
rs773891470 Chr.12: 3280342-3280342 on Build GRCh38
rs193079570 Chr.12: 3280535-3280535 on Build GRCh38
rs200763411 Chr.12: 3280529-3280529 on Build GRCh38
rs877089 Chr.12: 3280393-3280393 on Build GRCh38
rs1009404391 Chr.12: 3280395-3280395 on Build GRCh38
rs756939766 Chr.12: 3280502-3280502 on Build GRCh38
rs201570884 Chr.12: 3280339-3280339 on Build GRCh38
rs760396988 Chr.12: 3280436-3280436 on Build GRCh38
rs756396935 Chr.12: 3280394-3280394 on Build GRCh38
rs878983184 Chr.12: 3280405-3280405 on Build GRCh38
rs773599510 Chr.12: 3280445-3280445 on Build GRCh38
rs768231852 Chr.12: 3280523-3280523 on Build GRCh38
rs774771969 Chr.12: 3280488-3280488 on Build GRCh38
rs543141675 Chr.12: 3280345-3280345 on Build GRCh38
rs565829152 Chr.12: 3280339-3280340 on Build GRCh38
rs770783625 Chr.12: 3280423-3280423 on Build GRCh38
rs373609821 Chr.12: 3280497-3280497 on Build GRCh38
rs71577834 Chr.12: 3280372-3280372 on Build GRCh38
rs766796551 Chr.12: 3280459-3280459 on Build GRCh38
rs1040374613 Chr.12: 3280487-3280487 on Build GRCh38
rs151039760 Chr.12: 3280332-3280332 on Build GRCh38
rs35155720 Chr.12: 3280424-3280424 on Build GRCh38
rs375114637 Chr.12: 3280522-3280522 on Build GRCh38
rs775219139 Chr.12: 3280355-3280355 on Build GRCh38
rs377525804 Chr.12: 3280333-3280333 on Build GRCh38
rs763481991 Chr.12: 3280373-3280373 on Build GRCh38
rs745809480 Chr.12: 3280433-3280433 on Build GRCh38
rs201496490 Chr.12: 3280353-3280353 on Build GRCh38
rs745791104 Chr.12: 3280518-3280518 on Build GRCh38
rs756536556 Chr.12: 3280333-3280335 on Build GRCh38
rs745629435 Chr.12: 3280387-3280387 on Build GRCh38
rs748399308 Chr.12: 3280338-3280338 on Build GRCh38
rs763175388 Chr.12: 3280450-3280450 on Build GRCh38
rs749085831 Chr.12: 3280399-3280399 on Build GRCh38

View Full Product Details