Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6210214-6210447 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF207
Entrez Gene ID
388591
Gene Name
ring finger protein 207
Gene Aliases
C1orf188
UniGene
Hs.716549
Gene Chromosome Location
Chr.1: 6205475-6221299 on Build GRCh38
Transcripts
NM_207396.2 XM_011541439.2 XM_017001259.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7198596 c.873+1G>A p.? Chr.1: 6210296-6210296 on Build GRCh38
6362141 c.829A>G p.K277E Chr.1: 6210251-6210251 on Build GRCh38
1470349 c.801C>G p.S267R Chr.1: 6210223-6210223 on Build GRCh38
4722080 c.838_839delCT p.H282fs*26 Chr.1: 6210260-6210261 on Build GRCh38
2238130 c.801-8T>C p.? Chr.1: 6210215-6210215 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs140447252 Chr.1: 6210280-6210280 on Build GRCh38
rs772305460 Chr.1: 6210368-6210368 on Build GRCh38
rs765618061 Chr.1: 6210423-6210423 on Build GRCh38
rs754362484 Chr.1: 6210296-6210296 on Build GRCh38
rs541309541 Chr.1: 6210363-6210363 on Build GRCh38
rs148563561 Chr.1: 6210246-6210246 on Build GRCh38
rs754092948 Chr.1: 6210411-6210411 on Build GRCh38
rs377205229 Chr.1: 6210316-6210316 on Build GRCh38
rs767660036 Chr.1: 6210256-6210256 on Build GRCh38
rs113445340 Chr.1: 6210408-6210408 on Build GRCh38
rs779682896 Chr.1: 6210324-6210325 on Build GRCh38
rs761093083 Chr.1: 6210397-6210397 on Build GRCh38
rs761843305 Chr.1: 6210363-6210363 on Build GRCh38
rs12074555 Chr.1: 6210326-6210326 on Build GRCh38
rs531861750 Chr.1: 6210387-6210387 on Build GRCh38
rs201775287 Chr.1: 6210315-6210315 on Build GRCh38
rs543601566 Chr.1: 6210409-6210409 on Build GRCh38
rs769561399 Chr.1: 6210338-6210338 on Build GRCh38
rs745808987 Chr.1: 6210337-6210337 on Build GRCh38
rs1020193889 Chr.1: 6210328-6210328 on Build GRCh38
rs755988615 Chr.1: 6210284-6210287 on Build GRCh38
rs760089968 Chr.1: 6210416-6210416 on Build GRCh38
rs762037320 Chr.1: 6210247-6210247 on Build GRCh38
rs544727720 Chr.1: 6210244-6210244 on Build GRCh38
rs753629592 Chr.1: 6210356-6210357 on Build GRCh38
rs377158308 Chr.1: 6210367-6210367 on Build GRCh38
rs374550573 Chr.1: 6210275-6210275 on Build GRCh38
rs762985423 Chr.1: 6210243-6210243 on Build GRCh38
rs142653949 Chr.1: 6210343-6210343 on Build GRCh38
rs200803461 Chr.1: 6210233-6210233 on Build GRCh38
rs376517020 Chr.1: 6210308-6210308 on Build GRCh38
rs754503878 Chr.1: 6210303-6210303 on Build GRCh38
rs777610932 Chr.1: 6210427-6210427 on Build GRCh38
rs760980576 Chr.1: 6210269-6210269 on Build GRCh38
rs144299459 Chr.1: 6210425-6210425 on Build GRCh38
rs199849408 Chr.1: 6210309-6210309 on Build GRCh38
rs774419104 Chr.1: 6210355-6210355 on Build GRCh38
rs976051694 Chr.1: 6210370-6210370 on Build GRCh38
rs757956687 Chr.1: 6210426-6210426 on Build GRCh38
rs750819193 Chr.1: 6210266-6210266 on Build GRCh38
rs371571805 Chr.1: 6210305-6210305 on Build GRCh38

View Full Product Details