Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38313365-38313604 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC22A14
Entrez Gene ID
9389
Gene Name
solute carrier family 22 member 14
Gene Aliases
OCTL2, OCTL4, ORCTL4
UniGene
Hs.165559
Gene Chromosome Location
Chr.3: 38281662-38318575 on Build GRCh38
Transcripts
NM_001320033.1 NM_004803.4 XM_005265585.4 XM_006713416.3 XM_006713417.3 XM_006713418.3 XM_011534245.2 XM_011534247.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7164800 c.1122T>C p.N374N Chr.3: 38313444-38313444 on Build GRCh38
4836113 c.1131C>G p.L377L Chr.3: 38313453-38313453 on Build GRCh38
6317945 c.1066-9G>T p.? Chr.3: 38313379-38313379 on Build GRCh38
323370 c.1102G>T p.V368F Chr.3: 38313424-38313424 on Build GRCh38
6370637 c.1069C>A p.Q357K Chr.3: 38313391-38313391 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754331881 Chr.3: 38313416-38313416 on Build GRCh38
rs371034387 Chr.3: 38313525-38313525 on Build GRCh38
rs377493770 Chr.3: 38313532-38313532 on Build GRCh38
rs868692391 Chr.3: 38313551-38313551 on Build GRCh38
rs778636799 Chr.3: 38313503-38313503 on Build GRCh38
rs369861143 Chr.3: 38313402-38313402 on Build GRCh38
rs559722711 Chr.3: 38313517-38313517 on Build GRCh38
rs765525615 Chr.3: 38313443-38313443 on Build GRCh38
rs763383009 Chr.3: 38313533-38313533 on Build GRCh38
rs746283515 Chr.3: 38313442-38313443 on Build GRCh38
rs1019413477 Chr.3: 38313394-38313394 on Build GRCh38
rs934383123 Chr.3: 38313566-38313566 on Build GRCh38
rs1021066011 Chr.3: 38313490-38313490 on Build GRCh38
rs750814218 Chr.3: 38313445-38313445 on Build GRCh38
rs374194862 Chr.3: 38313528-38313528 on Build GRCh38
rs771760014 Chr.3: 38313512-38313512 on Build GRCh38
rs781286436 Chr.3: 38313477-38313477 on Build GRCh38
rs766814392 Chr.3: 38313537-38313537 on Build GRCh38
rs751733052 Chr.3: 38313459-38313459 on Build GRCh38
rs758751338 Chr.3: 38313451-38313451 on Build GRCh38
rs374014860 Chr.3: 38313444-38313444 on Build GRCh38
rs376693746 Chr.3: 38313502-38313502 on Build GRCh38
rs749193224 Chr.3: 38313494-38313494 on Build GRCh38
rs776142578 Chr.3: 38313527-38313527 on Build GRCh38
rs745644814 Chr.3: 38313509-38313509 on Build GRCh38
rs369257841 Chr.3: 38313396-38313396 on Build GRCh38
rs769369807 Chr.3: 38313476-38313476 on Build GRCh38
rs747949499 Chr.3: 38313411-38313411 on Build GRCh38
rs931411022 Chr.3: 38313540-38313540 on Build GRCh38
rs200813418 Chr.3: 38313415-38313415 on Build GRCh38
rs139343699 Chr.3: 38313468-38313468 on Build GRCh38
rs772301774 Chr.3: 38313478-38313479 on Build GRCh38
rs1047044120 Chr.3: 38313554-38313554 on Build GRCh38
rs746623147 Chr.3: 38313523-38313523 on Build GRCh38
rs775038075 Chr.3: 38313409-38313409 on Build GRCh38
rs780405742 Chr.3: 38313457-38313457 on Build GRCh38
rs762325578 Chr.3: 38313417-38313417 on Build GRCh38

View Full Product Details