Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 3292233-3292480 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
OR3A1
Entrez Gene ID
4994
Gene Name
olfactory receptor family 3 subfamily A member 1
Gene Aliases
OLFRA03, OR17-40, OR17-82, OR40
UniGene
Hs.647410
Gene Chromosome Location
Chr.17: 3288318-3336087 on Build GRCh38
Transcripts
NM_002550.2 XM_017024695.1 XM_017024696.1 XM_017024697.1 XM_017024698.1

Gene Symbol
OR3A2
Entrez Gene ID
4995
Gene Name
olfactory receptor family 3 subfamily A member 2
Gene Aliases
OLFRA04, OR17-14, OR17-228, OR228
UniGene
Hs.647414
Gene Chromosome Location
Chr.17: 3276113-3386311 on Build GRCh38
Transcripts
NM_002550.2 XM_017024695.1 XM_017024696.1 XM_017024697.1 XM_017024698.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5636878 c.252G>A p.M84I Chr.17: 3292331-3292331 on Build GRCh38
3180749 c.190T>G p.F64V Chr.17: 3292393-3292393 on Build GRCh38
705572 c.183C>A p.P61P Chr.17: 3292400-3292400 on Build GRCh38
705573 c.263T>C p.L88P Chr.17: 3292320-3292320 on Build GRCh38
4472998 c.181C>T p.P61S Chr.17: 3292402-3292402 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756901542 Chr.17: 3292456-3292456 on Build GRCh38
rs756795869 Chr.17: 3292287-3292287 on Build GRCh38
rs7218125 Chr.17: 3292309-3292309 on Build GRCh38
rs766058905 Chr.17: 3292436-3292436 on Build GRCh38
rs376929342 Chr.17: 3292348-3292348 on Build GRCh38
rs753624530 Chr.17: 3292406-3292406 on Build GRCh38
rs138089982 Chr.17: 3292303-3292303 on Build GRCh38
rs769908833 Chr.17: 3292312-3292312 on Build GRCh38
rs758609296 Chr.17: 3292256-3292256 on Build GRCh38
rs375681384 Chr.17: 3292253-3292253 on Build GRCh38
rs149271851 Chr.17: 3292320-3292320 on Build GRCh38
rs754730963 Chr.17: 3292444-3292444 on Build GRCh38
rs752443246 Chr.17: 3292324-3292324 on Build GRCh38
rs201853018 Chr.17: 3292404-3292404 on Build GRCh38
rs777711411 Chr.17: 3292282-3292282 on Build GRCh38
rs746745432 Chr.17: 3292378-3292378 on Build GRCh38
rs201171625 Chr.17: 3292410-3292410 on Build GRCh38
rs751248394 Chr.17: 3292442-3292442 on Build GRCh38
rs759821854 Chr.17: 3292401-3292401 on Build GRCh38
rs372689671 Chr.17: 3292311-3292311 on Build GRCh38
rs943455092 Chr.17: 3292330-3292330 on Build GRCh38
rs369569545 Chr.17: 3292260-3292260 on Build GRCh38
rs144484709 Chr.17: 3292349-3292349 on Build GRCh38
rs369737085 Chr.17: 3292288-3292288 on Build GRCh38
rs530368229 Chr.17: 3292376-3292376 on Build GRCh38
rs758081375 Chr.17: 3292346-3292346 on Build GRCh38
rs770993472 Chr.17: 3292460-3292460 on Build GRCh38
rs111595550 Chr.17: 3292337-3292337 on Build GRCh38
rs765117564 Chr.17: 3292429-3292429 on Build GRCh38
rs765314124 Chr.17: 3292252-3292252 on Build GRCh38
rs1036239372 Chr.17: 3292279-3292279 on Build GRCh38
rs756006843 Chr.17: 3292270-3292270 on Build GRCh38
rs749325194 Chr.17: 3292459-3292459 on Build GRCh38
rs774561559 Chr.17: 3292394-3292394 on Build GRCh38
rs542218726 Chr.17: 3292258-3292258 on Build GRCh38
rs770247541 Chr.17: 3292358-3292358 on Build GRCh38
rs778158296 Chr.17: 3292457-3292457 on Build GRCh38
rs757980013 Chr.17: 3292322-3292323 on Build GRCh38
rs778463360 Chr.17: 3292338-3292338 on Build GRCh38
rs16952828 Chr.17: 3292351-3292351 on Build GRCh38
rs763954548 Chr.17: 3292333-3292333 on Build GRCh38
rs372996458 Chr.17: 3292274-3292274 on Build GRCh38
rs867670908 Chr.17: 3292356-3292356 on Build GRCh38
rs779561803 Chr.17: 3292377-3292377 on Build GRCh38
rs773305474 Chr.17: 3292360-3292360 on Build GRCh38
rs148768371 Chr.17: 3292449-3292449 on Build GRCh38
rs749939112 Chr.17: 3292342-3292342 on Build GRCh38
rs150533143 Chr.17: 3292306-3292306 on Build GRCh38
rs61736316 Chr.17: 3292305-3292305 on Build GRCh38
rs927788626 Chr.17: 3292363-3292363 on Build GRCh38
rs745520779 Chr.17: 3292292-3292292 on Build GRCh38
rs867378787 Chr.17: 3292439-3292439 on Build GRCh38
rs749925251 Chr.17: 3292262-3292264 on Build GRCh38
rs749896963 Chr.17: 3292434-3292434 on Build GRCh38
rs769142492 Chr.17: 3292350-3292350 on Build GRCh38
rs752531310 Chr.17: 3292259-3292259 on Build GRCh38
rs771085179 Chr.17: 3292378-3292378 on Build GRCh38
rs760477994 Chr.17: 3292325-3292325 on Build GRCh38
rs756598283 Chr.17: 3292451-3292451 on Build GRCh38
rs547307019 Chr.17: 3292399-3292399 on Build GRCh38
rs773218076 Chr.17: 3292314-3292314 on Build GRCh38
rs753287732 Chr.17: 3292285-3292285 on Build GRCh38
rs147802348 Chr.17: 3292293-3292293 on Build GRCh38
rs748670575 Chr.17: 3292354-3292354 on Build GRCh38
rs761776576 Chr.17: 3292425-3292425 on Build GRCh38
rs146171012 Chr.17: 3292323-3292323 on Build GRCh38

View Full Product Details