Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 12825587-12825860 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RTBDN
Entrez Gene ID
83546
Gene Name
retbindin
Gene Aliases
-
UniGene
Hs.21162
Gene Chromosome Location
Chr.19: 12825477-12835428 on Build GRCh38
Transcripts
NM_001080997.2 NM_001270440.1 NM_001270441.1 NM_001270442.1 NM_001270443.1 NM_001270444.1 NM_001270445.1 NM_031429.2 XM_005260089.1 XM_005260090.3 XM_011528343.1 XM_011528344.2 XM_011528345.1 XM_017027343.1 XM_017027344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3528847 c.703T>G p.S235A Chr.19: 12825789-12825789 on Build GRCh38
5665833 c.771C>G p.S257R Chr.19: 12825721-12825721 on Build GRCh38
5567842 c.753C>T p.S251S Chr.19: 12825739-12825739 on Build GRCh38
1242799 c.764G>A p.S255N Chr.19: 12825728-12825728 on Build GRCh38
7215973 c.689G>C p.G230A Chr.19: 12825803-12825803 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs978300496 Chr.19: 12825755-12825755 on Build GRCh38
rs776047538 Chr.19: 12825689-12825689 on Build GRCh38
rs747608081 Chr.19: 12825798-12825798 on Build GRCh38
rs770518446 Chr.19: 12825738-12825738 on Build GRCh38
rs765358490 Chr.19: 12825701-12825701 on Build GRCh38
rs1048273402 Chr.19: 12825650-12825650 on Build GRCh38
rs747897365 Chr.19: 12825722-12825722 on Build GRCh38
rs755881349 Chr.19: 12825780-12825780 on Build GRCh38
rs750494324 Chr.19: 12825704-12825704 on Build GRCh38
rs574468963 Chr.19: 12825651-12825651 on Build GRCh38
rs778227828 Chr.19: 12825794-12825794 on Build GRCh38
rs1044708688 Chr.19: 12825691-12825691 on Build GRCh38
rs748614938 Chr.19: 12825773-12825774 on Build GRCh38
rs929857273 Chr.19: 12825628-12825628 on Build GRCh38
rs762089247 Chr.19: 12825756-12825756 on Build GRCh38
rs762140060 Chr.19: 12825807-12825807 on Build GRCh38
rs564096036 Chr.19: 12825765-12825765 on Build GRCh38
rs372483632 Chr.19: 12825833-12825833 on Build GRCh38
rs781112164 Chr.19: 12825775-12825775 on Build GRCh38
rs751401201 Chr.19: 12825712-12825712 on Build GRCh38
rs530478616 Chr.19: 12825745-12825745 on Build GRCh38
rs540034595 Chr.19: 12825655-12825655 on Build GRCh38
rs764324477 Chr.19: 12825692-12825692 on Build GRCh38
rs11104 Chr.19: 12825766-12825766 on Build GRCh38
rs768106839 Chr.19: 12825685-12825685 on Build GRCh38
rs754094252 Chr.19: 12825694-12825694 on Build GRCh38
rs374249027 Chr.19: 12825732-12825732 on Build GRCh38
rs377339618 Chr.19: 12825647-12825647 on Build GRCh38
rs770091984 Chr.19: 12825818-12825818 on Build GRCh38
rs751531402 Chr.19: 12825768-12825768 on Build GRCh38
rs895497054 Chr.19: 12825623-12825623 on Build GRCh38
rs146539925 Chr.19: 12825786-12825786 on Build GRCh38
rs369175490 Chr.19: 12825718-12825718 on Build GRCh38
rs776845810 Chr.19: 12825751-12825751 on Build GRCh38
rs377305288 Chr.19: 12825739-12825739 on Build GRCh38
rs559419717 Chr.19: 12825619-12825619 on Build GRCh38
rs754861528 Chr.19: 12825770-12825770 on Build GRCh38
rs779537988 Chr.19: 12825724-12825725 on Build GRCh38
rs756128548 Chr.19: 12825783-12825783 on Build GRCh38
rs891615413 Chr.19: 12825742-12825742 on Build GRCh38
rs777089977 Chr.19: 12825805-12825805 on Build GRCh38
rs750547731 Chr.19: 12825759-12825759 on Build GRCh38
rs552362874 Chr.19: 12825750-12825750 on Build GRCh38
rs763105175 Chr.19: 12825836-12825836 on Build GRCh38
rs777465724 Chr.19: 12825730-12825730 on Build GRCh38
rs755612598 Chr.19: 12825666-12825666 on Build GRCh38
rs761018662 Chr.19: 12825747-12825747 on Build GRCh38
rs1040680210 Chr.19: 12825615-12825615 on Build GRCh38
rs769570211 Chr.19: 12825726-12825726 on Build GRCh38
rs28582260 Chr.19: 12825792-12825792 on Build GRCh38
rs752403593 Chr.19: 12825777-12825777 on Build GRCh38
rs897067577 Chr.19: 12825621-12825621 on Build GRCh38
rs779941098 Chr.19: 12825709-12825709 on Build GRCh38
rs13628 Chr.19: 12825803-12825803 on Build GRCh38
rs182508783 Chr.19: 12825705-12825705 on Build GRCh38
rs761882939 Chr.19: 12825700-12825700 on Build GRCh38
rs145740526 Chr.19: 12825767-12825767 on Build GRCh38

View Full Product Details