Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2460104-2460351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf59
Entrez Gene ID
80178
Gene Name
chromosome 16 open reading frame 59
Gene Aliases
-
UniGene
Hs.534491
Gene Chromosome Location
Chr.16: 2460111-2464963 on Build GRCh38
Transcripts
NM_025108.2 XM_011522667.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4639408 c.1A>G p.M1V Chr.16: 2460145-2460145 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs775428546 Chr.16: 2460239-2460239 on Build GRCh38
rs566662488 Chr.16: 2460192-2460192 on Build GRCh38
rs750683166 Chr.16: 2460300-2460300 on Build GRCh38
rs532983159 Chr.16: 2460134-2460134 on Build GRCh38
rs746297783 Chr.16: 2460142-2460142 on Build GRCh38
rs770667313 Chr.16: 2460186-2460186 on Build GRCh38
rs934889728 Chr.16: 2460296-2460296 on Build GRCh38
rs769454948 Chr.16: 2460166-2460166 on Build GRCh38
rs373669956 Chr.16: 2460129-2460129 on Build GRCh38
rs748863700 Chr.16: 2460185-2460185 on Build GRCh38
rs34863489 Chr.16: 2460235-2460236 on Build GRCh38
rs1020039641 Chr.16: 2460139-2460139 on Build GRCh38
rs759392771 Chr.16: 2460210-2460210 on Build GRCh38
rs916947221 Chr.16: 2460323-2460323 on Build GRCh38
rs758964864 Chr.16: 2460152-2460152 on Build GRCh38
rs949109371 Chr.16: 2460229-2460229 on Build GRCh38
rs948387117 Chr.16: 2460331-2460331 on Build GRCh38
rs186033526 Chr.16: 2460153-2460153 on Build GRCh38
rs965359424 Chr.16: 2460304-2460304 on Build GRCh38
rs947578382 Chr.16: 2460135-2460135 on Build GRCh38
rs764126748 Chr.16: 2460263-2460263 on Build GRCh38
rs978924630 Chr.16: 2460270-2460270 on Build GRCh38
rs952838342 Chr.16: 2460154-2460154 on Build GRCh38
rs975589667 Chr.16: 2460132-2460132 on Build GRCh38
rs1056701817 Chr.16: 2460147-2460147 on Build GRCh38
rs757404893 Chr.16: 2460282-2460282 on Build GRCh38
rs757536703 Chr.16: 2460140-2460140 on Build GRCh38
rs902293539 Chr.16: 2460256-2460256 on Build GRCh38
rs767384204 Chr.16: 2460233-2460233 on Build GRCh38
rs917602341 Chr.16: 2460227-2460228 on Build GRCh38
rs541828211 Chr.16: 2460141-2460141 on Build GRCh38
rs895571533 Chr.16: 2460149-2460149 on Build GRCh38
rs747607391 Chr.16: 2460164-2460164 on Build GRCh38
rs1019595568 Chr.16: 2460207-2460207 on Build GRCh38
rs555677775 Chr.16: 2460268-2460268 on Build GRCh38
rs1019587244 Chr.16: 2460303-2460303 on Build GRCh38
rs765557858 Chr.16: 2460286-2460286 on Build GRCh38
rs867690350 Chr.16: 2460146-2460146 on Build GRCh38
rs535794554 Chr.16: 2460266-2460266 on Build GRCh38
rs994660948 Chr.16: 2460184-2460184 on Build GRCh38
rs965364502 Chr.16: 2460211-2460211 on Build GRCh38
rs1004220943 Chr.16: 2460195-2460195 on Build GRCh38
rs976295654 Chr.16: 2460196-2460196 on Build GRCh38
rs760704730 Chr.16: 2460261-2460261 on Build GRCh38
rs772882650 Chr.16: 2460177-2460177 on Build GRCh38

View Full Product Details