Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 150182711-150182964 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDX1
Entrez Gene ID
1044
Gene Name
caudal type homeobox 1
Gene Aliases
-
UniGene
Hs.1545
Gene Chromosome Location
Chr.5: 150166781-150184558 on Build GRCh38
Transcripts
NM_001804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4798571 c.546G>A p.R182R Chr.5: 150182868-150182868 on Build GRCh38
482341 c.496C>A p.R166S Chr.5: 150182818-150182818 on Build GRCh38
3212120 c.531T>C p.R177R Chr.5: 150182853-150182853 on Build GRCh38
4671286 c.473G>A p.R158H Chr.5: 150182795-150182795 on Build GRCh38
374522 c.529C>T p.R177C Chr.5: 150182851-150182851 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200074812 Chr.5: 150182852-150182852 on Build GRCh38
rs746782770 Chr.5: 150182825-150182825 on Build GRCh38
rs143202475 Chr.5: 150182908-150182908 on Build GRCh38
rs771838490 Chr.5: 150182754-150182754 on Build GRCh38
rs545571678 Chr.5: 150182867-150182867 on Build GRCh38
rs562358188 Chr.5: 150182901-150182901 on Build GRCh38
rs770561349 Chr.5: 150182795-150182795 on Build GRCh38
rs768382207 Chr.5: 150182897-150182897 on Build GRCh38
rs760129484 Chr.5: 150182917-150182917 on Build GRCh38
rs775151093 Chr.5: 150182761-150182761 on Build GRCh38
rs774953728 Chr.5: 150182916-150182916 on Build GRCh38
rs1055732427 Chr.5: 150182763-150182763 on Build GRCh38
rs781624212 Chr.5: 150182741-150182741 on Build GRCh38
rs753003920 Chr.5: 150182808-150182808 on Build GRCh38
rs760527166 Chr.5: 150182768-150182768 on Build GRCh38
rs776226683 Chr.5: 150182843-150182843 on Build GRCh38
rs764675920 Chr.5: 150182780-150182780 on Build GRCh38
rs1032049779 Chr.5: 150182898-150182898 on Build GRCh38
rs771897143 Chr.5: 150182818-150182818 on Build GRCh38
rs556449017 Chr.5: 150182777-150182777 on Build GRCh38
rs140956520 Chr.5: 150182869-150182869 on Build GRCh38
rs781538102 Chr.5: 150182807-150182807 on Build GRCh38
rs761281057 Chr.5: 150182779-150182779 on Build GRCh38
rs772595006 Chr.5: 150182848-150182848 on Build GRCh38
rs761468699 Chr.5: 150182846-150182846 on Build GRCh38
rs770509761 Chr.5: 150182915-150182915 on Build GRCh38
rs146129317 Chr.5: 150182866-150182866 on Build GRCh38
rs745812552 Chr.5: 150182753-150182753 on Build GRCh38
rs768479920 Chr.5: 150182835-150182835 on Build GRCh38
rs1007473110 Chr.5: 150182889-150182889 on Build GRCh38
rs143926295 Chr.5: 150182847-150182847 on Build GRCh38
rs750884828 Chr.5: 150182783-150182783 on Build GRCh38
rs184290965 Chr.5: 150182883-150182883 on Build GRCh38
rs199986969 Chr.5: 150182748-150182748 on Build GRCh38
rs756165293 Chr.5: 150182809-150182809 on Build GRCh38
rs746978526 Chr.5: 150182891-150182891 on Build GRCh38
rs200155434 Chr.5: 150182870-150182870 on Build GRCh38
rs762583452 Chr.5: 150182850-150182850 on Build GRCh38
rs200303526 Chr.5: 150182939-150182939 on Build GRCh38
rs745748042 Chr.5: 150182813-150182813 on Build GRCh38
rs762341206 Chr.5: 150182912-150182912 on Build GRCh38
rs150404926 Chr.5: 150182796-150182796 on Build GRCh38
rs548360415 Chr.5: 150182797-150182797 on Build GRCh38
rs757405244 Chr.5: 150182875-150182875 on Build GRCh38
rs910011329 Chr.5: 150182769-150182769 on Build GRCh38
rs373496881 Chr.5: 150182846-150182847 on Build GRCh38
rs141296812 Chr.5: 150182782-150182782 on Build GRCh38
rs138123617 Chr.5: 150182798-150182798 on Build GRCh38
rs201857355 Chr.5: 150182851-150182851 on Build GRCh38
rs574805115 Chr.5: 150182824-150182824 on Build GRCh38
rs770039074 Chr.5: 150182749-150182749 on Build GRCh38
rs767922855 Chr.5: 150182918-150182918 on Build GRCh38
rs769319816 Chr.5: 150182909-150182909 on Build GRCh38

View Full Product Details