Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 137225703-137225934 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYSRT1
Entrez Gene ID
375791
Gene Name
cysteine rich tail 1
Gene Aliases
C9orf169
UniGene
Hs.495540
Gene Chromosome Location
Chr.9: 137224635-137226311 on Build GRCh38
Transcripts
NM_199001.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1199084 c.145C>T p.P49S Chr.9: 137225766-137225766 on Build GRCh38
4794923 c.278G>A p.S93N Chr.9: 137225899-137225899 on Build GRCh38
7237468 c.145C>A p.P49T Chr.9: 137225766-137225766 on Build GRCh38
4645673 c.287G>A p.R96H Chr.9: 137225908-137225908 on Build GRCh38
4632497 c.163A>G p.T55A Chr.9: 137225784-137225784 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs546990293 Chr.9: 137225877-137225877 on Build GRCh38
rs766411779 Chr.9: 137225825-137225825 on Build GRCh38
rs775766198 Chr.9: 137225786-137225786 on Build GRCh38
rs749958563 Chr.9: 137225808-137225808 on Build GRCh38
rs745952298 Chr.9: 137225916-137225916 on Build GRCh38
rs530388424 Chr.9: 137225870-137225870 on Build GRCh38
rs1038295886 Chr.9: 137225766-137225767 on Build GRCh38
rs764579844 Chr.9: 137225801-137225801 on Build GRCh38
rs773856592 Chr.9: 137225803-137225803 on Build GRCh38
rs941023819 Chr.9: 137225750-137225750 on Build GRCh38
rs763305937 Chr.9: 137225790-137225790 on Build GRCh38
rs939743872 Chr.9: 137225731-137225731 on Build GRCh38
rs146301609 Chr.9: 137225869-137225869 on Build GRCh38
rs757522242 Chr.9: 137225886-137225886 on Build GRCh38
rs1045565163 Chr.9: 137225811-137225811 on Build GRCh38
rs955867056 Chr.9: 137225800-137225800 on Build GRCh38
rs957432866 Chr.9: 137225853-137225853 on Build GRCh38
rs777169250 Chr.9: 137225741-137225741 on Build GRCh38
rs531075325 Chr.9: 137225805-137225805 on Build GRCh38
rs141991047 Chr.9: 137225824-137225824 on Build GRCh38
rs770200987 Chr.9: 137225826-137225826 on Build GRCh38
rs973845687 Chr.9: 137225900-137225900 on Build GRCh38
rs776805165 Chr.9: 137225744-137225744 on Build GRCh38
rs1031481474 Chr.9: 137225792-137225792 on Build GRCh38
rs761570151 Chr.9: 137225804-137225804 on Build GRCh38
rs182088466 Chr.9: 137225736-137225736 on Build GRCh38
rs532569400 Chr.9: 137225908-137225908 on Build GRCh38
rs1032164357 Chr.9: 137225834-137225834 on Build GRCh38
rs983681320 Chr.9: 137225847-137225847 on Build GRCh38
rs567013326 Chr.9: 137225885-137225885 on Build GRCh38
rs960256551 Chr.9: 137225840-137225840 on Build GRCh38
rs746326548 Chr.9: 137225785-137225785 on Build GRCh38
rs552400420 Chr.9: 137225914-137225914 on Build GRCh38

View Full Product Details