Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 77920218-77920455 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHTF2
Entrez Gene ID
57157
Gene Name
putative homeodomain transcription factor 2
Gene Aliases
-
UniGene
Hs.203965
Gene Chromosome Location
Chr.7: 77798792-77957504 on Build GRCh38
Transcripts
NM_001127357.1 NM_001127358.1 NM_001127359.1 NM_001127360.1 NM_020432.4 XM_005250508.4 XM_005250509.4 XM_006716063.3 XM_011516422.2 XM_011516423.2 XM_011516424.2 XM_017012458.1 XM_017012459.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1091963 c.828C>A p.G276G Chr.7: 77920330-77920330 on Build GRCh38
1091961 c.792G>T p.K264N Chr.7: 77920294-77920294 on Build GRCh38
1091959 c.783G>T p.K261N Chr.7: 77920285-77920285 on Build GRCh38
3882516 c.735C>T p.S245S Chr.7: 77920339-77920339 on Build GRCh38
6548377 c.687A>T p.A229A Chr.7: 77920291-77920291 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763675811 Chr.7: 77920364-77920364 on Build GRCh38
rs940428856 Chr.7: 77920333-77920333 on Build GRCh38
rs201907528 Chr.7: 77920356-77920356 on Build GRCh38
rs375949234 Chr.7: 77920367-77920367 on Build GRCh38
rs1034305054 Chr.7: 77920358-77920358 on Build GRCh38
rs367743132 Chr.7: 77920383-77920383 on Build GRCh38
rs780362374 Chr.7: 77920381-77920381 on Build GRCh38
rs368157181 Chr.7: 77920413-77920413 on Build GRCh38
rs897425531 Chr.7: 77920289-77920289 on Build GRCh38
rs758950839 Chr.7: 77920370-77920370 on Build GRCh38
rs749971460 Chr.7: 77920427-77920427 on Build GRCh38
rs767408880 Chr.7: 77920341-77920341 on Build GRCh38
rs763733718 Chr.7: 77920409-77920409 on Build GRCh38
rs570568488 Chr.7: 77920407-77920407 on Build GRCh38
rs371277530 Chr.7: 77920366-77920366 on Build GRCh38
rs202151098 Chr.7: 77920252-77920252 on Build GRCh38
rs1045402380 Chr.7: 77920338-77920338 on Build GRCh38
rs371306987 Chr.7: 77920384-77920384 on Build GRCh38
rs886149918 Chr.7: 77920324-77920324 on Build GRCh38
rs922079045 Chr.7: 77920253-77920253 on Build GRCh38
rs759615826 Chr.7: 77920294-77920294 on Build GRCh38
rs773401003 Chr.7: 77920389-77920389 on Build GRCh38
rs745779028 Chr.7: 77920397-77920397 on Build GRCh38
rs374229433 Chr.7: 77920387-77920387 on Build GRCh38
rs772176287 Chr.7: 77920398-77920398 on Build GRCh38
rs377646178 Chr.7: 77920248-77920248 on Build GRCh38
rs775475306 Chr.7: 77920354-77920354 on Build GRCh38
rs769762092 Chr.7: 77920388-77920388 on Build GRCh38
rs563012048 Chr.7: 77920257-77920257 on Build GRCh38
rs761582191 Chr.7: 77920425-77920425 on Build GRCh38
rs764627313 Chr.7: 77920426-77920426 on Build GRCh38
rs764866568 Chr.7: 77920368-77920368 on Build GRCh38
rs775375328 Chr.7: 77920400-77920400 on Build GRCh38
rs781287451 Chr.7: 77920386-77920386 on Build GRCh38
rs371590586 Chr.7: 77920285-77920285 on Build GRCh38
rs906892872 Chr.7: 77920343-77920343 on Build GRCh38
rs748203544 Chr.7: 77920265-77920265 on Build GRCh38
rs774163024 Chr.7: 77920286-77920286 on Build GRCh38

View Full Product Details