Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72202580-72202811 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1109966 c.230G>A p.G77E Chr.9: 72202588-72202588 on Build GRCh38
4432417 c.251A>G p.H84R Chr.9: 72202609-72202609 on Build GRCh38
5890420 c.325C>T p.P109S Chr.9: 72202683-72202683 on Build GRCh38
4416937 c.326C>T p.P109L Chr.9: 72202684-72202684 on Build GRCh38
4611518 c.360_362delAGA p.E122delE Chr.9: 72202718-72202720 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766142263 Chr.9: 72202764-72202769 on Build GRCh38
rs763084635 Chr.9: 72202657-72202657 on Build GRCh38
rs529545864 Chr.9: 72202604-72202604 on Build GRCh38
rs755331133 Chr.9: 72202610-72202610 on Build GRCh38
rs773222504 Chr.9: 72202653-72202653 on Build GRCh38
rs149388704 Chr.9: 72202761-72202761 on Build GRCh38
rs1031238767 Chr.9: 72202710-72202710 on Build GRCh38
rs761091832 Chr.9: 72202774-72202774 on Build GRCh38
rs762914194 Chr.9: 72202683-72202683 on Build GRCh38
rs762960160 Chr.9: 72202759-72202759 on Build GRCh38
rs747963673 Chr.9: 72202752-72202752 on Build GRCh38
rs75382038 Chr.9: 72202758-72202758 on Build GRCh38
rs779979867 Chr.9: 72202715-72202715 on Build GRCh38
rs371223443 Chr.9: 72202659-72202659 on Build GRCh38
rs779961418 Chr.9: 72202644-72202644 on Build GRCh38
rs768112787 Chr.9: 72202767-72202767 on Build GRCh38
rs745358144 Chr.9: 72202642-72202642 on Build GRCh38
rs985092341 Chr.9: 72202769-72202769 on Build GRCh38
rs146422167 Chr.9: 72202651-72202651 on Build GRCh38
rs764800573 Chr.9: 72202775-72202775 on Build GRCh38
rs771958358 Chr.9: 72202643-72202643 on Build GRCh38
rs753618221 Chr.9: 72202778-72202781 on Build GRCh38
rs113810444 Chr.9: 72202731-72202731 on Build GRCh38
rs1052956672 Chr.9: 72202769-72202791 on Build GRCh38
rs530521050 Chr.9: 72202677-72202677 on Build GRCh38
rs750356146 Chr.9: 72202756-72202757 on Build GRCh38
rs753494302 Chr.9: 72202633-72202633 on Build GRCh38
rs756826268 Chr.9: 72202640-72202640 on Build GRCh38
rs781016341 Chr.9: 72202741-72202741 on Build GRCh38
rs975141723 Chr.9: 72202757-72202757 on Build GRCh38
rs775031438 Chr.9: 72202762-72202762 on Build GRCh38
rs138792393 Chr.9: 72202678-72202678 on Build GRCh38
rs746898915 Chr.9: 72202727-72202727 on Build GRCh38
rs776859539 Chr.9: 72202646-72202646 on Build GRCh38
rs753059474 Chr.9: 72202768-72202768 on Build GRCh38
rs956509172 Chr.9: 72202720-72202720 on Build GRCh38
rs767384214 Chr.9: 72202718-72202720 on Build GRCh38
rs778323963 Chr.9: 72202641-72202641 on Build GRCh38
rs777586157 Chr.9: 72202613-72202613 on Build GRCh38
rs767957340 Chr.9: 72202673-72202673 on Build GRCh38
rs749965441 Chr.9: 72202707-72202707 on Build GRCh38
rs371294481 Chr.9: 72202754-72202754 on Build GRCh38
rs767002186 Chr.9: 72202668-72202668 on Build GRCh38
rs778610772 Chr.9: 72202700-72202700 on Build GRCh38
rs1018944322 Chr.9: 72202753-72202753 on Build GRCh38
rs61758972 Chr.9: 72202670-72202670 on Build GRCh38
rs761986169 Chr.9: 72202709-72202709 on Build GRCh38
rs77548027 Chr.9: 72202645-72202645 on Build GRCh38
rs368070896 Chr.9: 72202730-72202730 on Build GRCh38
rs768469943 Chr.9: 72202736-72202736 on Build GRCh38
rs760664067 Chr.9: 72202759-72202760 on Build GRCh38

View Full Product Details