Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 144742-145015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLEKHG4B
Entrez Gene ID
153478
Gene Name
pleckstrin homology and RhoGEF domain containing G4B
Gene Aliases
-
UniGene
Hs.535800
Gene Chromosome Location
Chr.5: 54065-189972 on Build GRCh38
Transcripts
NM_052909.3 XM_006714446.3 XM_011513968.2 XM_011513969.2 XM_011513970.1 XM_011513971.2 XM_011513972.2 XM_011513973.1 XM_017009112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3674341 c.755G>T p.R252L Chr.5: 144838-144838 on Build GRCh38
6221840 c.837+5G>A p.? Chr.5: 144925-144925 on Build GRCh38
1063727 c.791G>A p.R264K Chr.5: 144874-144874 on Build GRCh38
4559804 c.819G>A p.Q273Q Chr.5: 144902-144902 on Build GRCh38
6348736 c.808G>T p.A270S Chr.5: 144891-144891 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777340824 Chr.5: 144926-144926 on Build GRCh38
rs757941005 Chr.5: 144885-144885 on Build GRCh38
rs373901696 Chr.5: 144969-144969 on Build GRCh38
rs1003974438 Chr.5: 144981-144981 on Build GRCh38
rs936918212 Chr.5: 144775-144775 on Build GRCh38
rs765743785 Chr.5: 144937-144937 on Build GRCh38
rs147627379 Chr.5: 144962-144962 on Build GRCh38
rs1015003553 Chr.5: 144994-144994 on Build GRCh38
rs985683051 Chr.5: 144809-144809 on Build GRCh38
rs530587991 Chr.5: 144824-144824 on Build GRCh38
rs766983614 Chr.5: 144961-144961 on Build GRCh38
rs754513050 Chr.5: 144958-144958 on Build GRCh38
rs765893879 Chr.5: 144952-144952 on Build GRCh38
rs973219106 Chr.5: 144765-144765 on Build GRCh38
rs761273749 Chr.5: 144858-144858 on Build GRCh38
rs759725521 Chr.5: 144852-144852 on Build GRCh38
rs748074396 Chr.5: 144894-144894 on Build GRCh38
rs920051385 Chr.5: 144769-144769 on Build GRCh38
rs1053745297 Chr.5: 144798-144798 on Build GRCh38
rs945492540 Chr.5: 144836-144836 on Build GRCh38
rs202227892 Chr.5: 144837-144837 on Build GRCh38
rs190985509 Chr.5: 144776-144776 on Build GRCh38
rs567687691 Chr.5: 144867-144867 on Build GRCh38
rs755945244 Chr.5: 144899-144899 on Build GRCh38
rs970501923 Chr.5: 144984-144984 on Build GRCh38
rs773674170 Chr.5: 144942-144942 on Build GRCh38
rs777148667 Chr.5: 144861-144861 on Build GRCh38
rs750971693 Chr.5: 144888-144888 on Build GRCh38
rs777574312 Chr.5: 144966-144966 on Build GRCh38
rs557331812 Chr.5: 144982-144982 on Build GRCh38
rs746991216 Chr.5: 144788-144788 on Build GRCh38
rs373035855 Chr.5: 144893-144893 on Build GRCh38
rs142186656 Chr.5: 144870-144870 on Build GRCh38
rs757742642 Chr.5: 144948-144948 on Build GRCh38
rs548182920 Chr.5: 144770-144770 on Build GRCh38
rs568462384 Chr.5: 144924-144924 on Build GRCh38
rs187321186 Chr.5: 144992-144992 on Build GRCh38
rs769563234 Chr.5: 144838-144838 on Build GRCh38
rs980941560 Chr.5: 144995-144995 on Build GRCh38
rs146958613 Chr.5: 144828-144828 on Build GRCh38
rs377204835 Chr.5: 144970-144970 on Build GRCh38
rs768763317 Chr.5: 144925-144925 on Build GRCh38
rs147604765 Chr.5: 144859-144859 on Build GRCh38
rs547156855 Chr.5: 144909-144909 on Build GRCh38
rs762675095 Chr.5: 144945-144945 on Build GRCh38
rs1008516071 Chr.5: 144891-144891 on Build GRCh38
rs751216322 Chr.5: 144957-144957 on Build GRCh38
rs762321297 Chr.5: 144863-144863 on Build GRCh38
rs762376224 Chr.5: 144931-144931 on Build GRCh38
rs183394750 Chr.5: 144988-144988 on Build GRCh38
rs775380932 Chr.5: 144855-144855 on Build GRCh38
rs746108293 Chr.5: 144904-144904 on Build GRCh38
rs774623082 Chr.5: 144843-144843 on Build GRCh38
rs570690532 Chr.5: 144865-144865 on Build GRCh38
rs765949040 Chr.5: 144887-144887 on Build GRCh38
rs367577014 Chr.5: 144889-144889 on Build GRCh38

View Full Product Details