Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 156963852-156964093 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NOM1
Entrez Gene ID
64434
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
C7orf3, PPP1R113, SGD1
UniGene
Hs.15825
Gene Chromosome Location
Chr.7: 156949723-156973182 on Build GRCh38
Transcripts
NM_138400.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7076885 c.1923G>C p.K641N Chr.7: 156963916-156963916 on Build GRCh38
3305843 c.2030T>C p.L677P Chr.7: 156964023-156964023 on Build GRCh38
1755187 c.1919C>T p.S640L Chr.7: 156963912-156963912 on Build GRCh38
1087901 c.1952T>C p.M651T Chr.7: 156963945-156963945 on Build GRCh38
3358113 c.1953G>A p.M651I Chr.7: 156963946-156963946 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369726557 Chr.7: 156963894-156963894 on Build GRCh38
rs747852442 Chr.7: 156964037-156964037 on Build GRCh38
rs375289491 Chr.7: 156963904-156963904 on Build GRCh38
rs775667123 Chr.7: 156963953-156963953 on Build GRCh38
rs374779806 Chr.7: 156963979-156963979 on Build GRCh38
rs765513172 Chr.7: 156963875-156963875 on Build GRCh38
rs192435513 Chr.7: 156963874-156963874 on Build GRCh38
rs756399939 Chr.7: 156963905-156963905 on Build GRCh38
rs753141852 Chr.7: 156963877-156963877 on Build GRCh38
rs114813001 Chr.7: 156963933-156963933 on Build GRCh38
rs114916388 Chr.7: 156963939-156963939 on Build GRCh38
rs752067057 Chr.7: 156963997-156963997 on Build GRCh38
rs372415851 Chr.7: 156963974-156963974 on Build GRCh38
rs749133837 Chr.7: 156963928-156963928 on Build GRCh38
rs758877378 Chr.7: 156963881-156963881 on Build GRCh38
rs10267297 Chr.7: 156963952-156963952 on Build GRCh38
rs756243178 Chr.7: 156963993-156963993 on Build GRCh38
rs376421854 Chr.7: 156963887-156963887 on Build GRCh38
rs757332240 Chr.7: 156963895-156963895 on Build GRCh38
rs764515530 Chr.7: 156963968-156963968 on Build GRCh38
rs200882197 Chr.7: 156963971-156963971 on Build GRCh38
rs753515293 Chr.7: 156964026-156964026 on Build GRCh38
rs763338650 Chr.7: 156963960-156963960 on Build GRCh38
rs752824373 Chr.7: 156963909-156963909 on Build GRCh38
rs997244263 Chr.7: 156963925-156963925 on Build GRCh38
rs113198844 Chr.7: 156963911-156963911 on Build GRCh38
rs750566378 Chr.7: 156963992-156963992 on Build GRCh38
rs779692904 Chr.7: 156963920-156963920 on Build GRCh38
rs748091585 Chr.7: 156964033-156964033 on Build GRCh38
rs149887319 Chr.7: 156963929-156963929 on Build GRCh38
rs780358064 Chr.7: 156964012-156964012 on Build GRCh38
rs754835639 Chr.7: 156964029-156964029 on Build GRCh38
rs752073441 Chr.7: 156963970-156963970 on Build GRCh38
rs199598246 Chr.7: 156963903-156963903 on Build GRCh38
rs369881632 Chr.7: 156963942-156963942 on Build GRCh38
rs750751219 Chr.7: 156963932-156963932 on Build GRCh38
rs772756564 Chr.7: 156963871-156963871 on Build GRCh38

View Full Product Details