Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33798540-33798777 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDCD6IP
Entrez Gene ID
10015
Gene Name
programmed cell death 6 interacting protein
Gene Aliases
AIP1, ALIX, DRIP4, HP95
UniGene
Hs.475896
Gene Chromosome Location
Chr.3: 33798571-33869707 on Build GRCh38
Transcripts
NM_001162429.2 NM_001256192.1 NM_013374.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
304121 c.34A>C p.T12P Chr.3: 33798762-33798762 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs879206808 Chr.3: 33798652-33798652 on Build GRCh38
rs986246057 Chr.3: 33798655-33798655 on Build GRCh38
rs552254221 Chr.3: 33798733-33798733 on Build GRCh38
rs550890558 Chr.3: 33798649-33798649 on Build GRCh38
rs28381976 Chr.3: 33798607-33798607 on Build GRCh38
rs1013370958 Chr.3: 33798634-33798634 on Build GRCh38
rs1141054 Chr.3: 33798622-33798622 on Build GRCh38
rs760977461 Chr.3: 33798746-33798746 on Build GRCh38
rs963733215 Chr.3: 33798606-33798606 on Build GRCh38
rs774852061 Chr.3: 33798709-33798711 on Build GRCh38
rs1048016998 Chr.3: 33798751-33798751 on Build GRCh38
rs11554560 Chr.3: 33798747-33798747 on Build GRCh38
rs993779340 Chr.3: 33798732-33798732 on Build GRCh38
rs917363843 Chr.3: 33798685-33798685 on Build GRCh38
rs1005629565 Chr.3: 33798597-33798597 on Build GRCh38
rs954484684 Chr.3: 33798653-33798653 on Build GRCh38
rs888412193 Chr.3: 33798640-33798640 on Build GRCh38
rs372627622 Chr.3: 33798651-33798652 on Build GRCh38
rs759061176 Chr.3: 33798713-33798713 on Build GRCh38
rs937682835 Chr.3: 33798595-33798595 on Build GRCh38
rs529896479 Chr.3: 33798656-33798656 on Build GRCh38
rs768111776 Chr.3: 33798646-33798646 on Build GRCh38
rs189555154 Chr.3: 33798660-33798660 on Build GRCh38
rs977397728 Chr.3: 33798629-33798629 on Build GRCh38
rs766391698 Chr.3: 33798660-33798663 on Build GRCh38
rs760193817 Chr.3: 33798632-33798633 on Build GRCh38
rs947622255 Chr.3: 33798598-33798598 on Build GRCh38
rs768705106 Chr.3: 33798682-33798682 on Build GRCh38
rs773719661 Chr.3: 33798705-33798705 on Build GRCh38
rs776725231 Chr.3: 33798683-33798683 on Build GRCh38
rs1057469789 Chr.3: 33798612-33798612 on Build GRCh38
rs371387868 Chr.3: 33798704-33798704 on Build GRCh38
rs1043296613 Chr.3: 33798603-33798603 on Build GRCh38
rs1054424067 Chr.3: 33798641-33798641 on Build GRCh38
rs924519844 Chr.3: 33798633-33798633 on Build GRCh38
rs866425760 Chr.3: 33798745-33798745 on Build GRCh38
rs968344942 Chr.3: 33798658-33798658 on Build GRCh38
rs376758894 Chr.3: 33798721-33798721 on Build GRCh38
rs368948546 Chr.3: 33798650-33798650 on Build GRCh38
rs747888688 Chr.3: 33798690-33798690 on Build GRCh38
rs1044404663 Chr.3: 33798638-33798638 on Build GRCh38
rs961821773 Chr.3: 33798578-33798578 on Build GRCh38
rs934797039 Chr.3: 33798611-33798611 on Build GRCh38
rs867033284 Chr.3: 33798740-33798740 on Build GRCh38
rs910847147 Chr.3: 33798596-33798596 on Build GRCh38
rs546293941 Chr.3: 33798587-33798587 on Build GRCh38
rs1006105985 Chr.3: 33798642-33798642 on Build GRCh38
rs1016644228 Chr.3: 33798600-33798600 on Build GRCh38
rs200697599 Chr.3: 33798742-33798742 on Build GRCh38
rs961878862 Chr.3: 33798648-33798648 on Build GRCh38
rs879131178 Chr.3: 33798662-33798662 on Build GRCh38
rs762492079 Chr.3: 33798697-33798697 on Build GRCh38
rs954557757 Chr.3: 33798590-33798590 on Build GRCh38
rs769744748 Chr.3: 33798692-33798692 on Build GRCh38
rs934857814 Chr.3: 33798664-33798664 on Build GRCh38
rs566691033 Chr.3: 33798693-33798693 on Build GRCh38
rs1016056780 Chr.3: 33798558-33798558 on Build GRCh38
rs993007185 Chr.3: 33798680-33798680 on Build GRCh38
rs890252819 Chr.3: 33798753-33798753 on Build GRCh38
rs972244967 Chr.3: 33798581-33798581 on Build GRCh38
rs985807766 Chr.3: 33798594-33798594 on Build GRCh38
rs1030168366 Chr.3: 33798743-33798743 on Build GRCh38
rs941348915 Chr.3: 33798700-33798700 on Build GRCh38

View Full Product Details