Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 63592482-63592721 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMEB2
Entrez Gene ID
26205
Gene Name
glucocorticoid modulatory element binding protein 2
Gene Aliases
GMEB-2, P79PIF, PIF79
UniGene
Hs.473286
Gene Chromosome Location
Chr.20: 63587594-63627101 on Build GRCh38
Transcripts
NM_012384.4 XM_005260202.4 XM_006723776.2 XM_011528779.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5343656 c.809A>T p.D270V Chr.20: 63592553-63592553 on Build GRCh38
4756348 c.761A>G p.H254R Chr.20: 63592601-63592601 on Build GRCh38
5707515 c.775G>A p.E259K Chr.20: 63592587-63592587 on Build GRCh38
3549108 c.804C>T p.V268V Chr.20: 63592558-63592558 on Build GRCh38
4903588 c.814C>T p.P272S Chr.20: 63592548-63592548 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766491192 Chr.20: 63592636-63592636 on Build GRCh38
rs550589899 Chr.20: 63592694-63592694 on Build GRCh38
rs368815293 Chr.20: 63592594-63592594 on Build GRCh38
rs750601250 Chr.20: 63592513-63592513 on Build GRCh38
rs142326335 Chr.20: 63592553-63592553 on Build GRCh38
rs751564384 Chr.20: 63592539-63592539 on Build GRCh38
rs755097129 Chr.20: 63592540-63592540 on Build GRCh38
rs893701289 Chr.20: 63592525-63592525 on Build GRCh38
rs779737136 Chr.20: 63592703-63592703 on Build GRCh38
rs778158894 Chr.20: 63592678-63592678 on Build GRCh38
rs768527058 Chr.20: 63592624-63592624 on Build GRCh38
rs371180146 Chr.20: 63592520-63592520 on Build GRCh38
rs770519690 Chr.20: 63592597-63592597 on Build GRCh38
rs151280948 Chr.20: 63592573-63592573 on Build GRCh38
rs1021276090 Chr.20: 63592677-63592695 on Build GRCh38
rs375401346 Chr.20: 63592578-63592578 on Build GRCh38
rs767584610 Chr.20: 63592660-63592660 on Build GRCh38
rs771775837 Chr.20: 63592617-63592617 on Build GRCh38
rs757687711 Chr.20: 63592686-63592686 on Build GRCh38
rs201344944 Chr.20: 63592515-63592515 on Build GRCh38
rs769306023 Chr.20: 63592535-63592535 on Build GRCh38
rs778012093 Chr.20: 63592582-63592582 on Build GRCh38
rs371080103 Chr.20: 63592702-63592702 on Build GRCh38
rs745359300 Chr.20: 63592608-63592608 on Build GRCh38
rs753266984 Chr.20: 63592565-63592565 on Build GRCh38
rs753140474 Chr.20: 63592675-63592675 on Build GRCh38
rs751863129 Chr.20: 63592639-63592639 on Build GRCh38
rs760702242 Chr.20: 63592621-63592621 on Build GRCh38
rs144182072 Chr.20: 63592635-63592635 on Build GRCh38
rs763107793 Chr.20: 63592530-63592530 on Build GRCh38
rs372554080 Chr.20: 63592620-63592620 on Build GRCh38
rs942739570 Chr.20: 63592547-63592547 on Build GRCh38
rs145518654 Chr.20: 63592633-63592633 on Build GRCh38
rs535253760 Chr.20: 63592645-63592645 on Build GRCh38
rs181561777 Chr.20: 63592677-63592677 on Build GRCh38
rs763990282 Chr.20: 63592511-63592511 on Build GRCh38
rs745521476 Chr.20: 63592696-63592696 on Build GRCh38
rs140508139 Chr.20: 63592600-63592600 on Build GRCh38
rs754119534 Chr.20: 63592685-63592685 on Build GRCh38

View Full Product Details