Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134566220-134566461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JADE2
Entrez Gene ID
23338
Gene Name
jade family PHD finger 2
Gene Aliases
JADE-2, PHF15
UniGene
Hs.732155
Gene Chromosome Location
Chr.5: 134524154-134583230 on Build GRCh38
Transcripts
NM_001289984.1 NM_001289985.1 NM_001308143.1 NM_015288.5 XM_005271945.1 XM_005271946.1 XM_005271948.4 XM_011543291.2 XM_017009282.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3760917 c.1151G>T p.G384V Chr.5: 134566249-134566249 on Build GRCh38
7157732 c.1234G>A p.E412K Chr.5: 134566380-134566380 on Build GRCh38
6316468 c.1272G>T p.V424V Chr.5: 134566370-134566370 on Build GRCh38
3138105 c.1177C>T p.R393C Chr.5: 134566323-134566323 on Build GRCh38
5834939 c.1229_1230insCGA p.P410_A411insE Chr.5: 134566375-134566376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs137957798 Chr.5: 134566254-134566254 on Build GRCh38
rs747052349 Chr.5: 134566279-134566279 on Build GRCh38
rs578254108 Chr.5: 134566243-134566243 on Build GRCh38
rs771026777 Chr.5: 134566425-134566425 on Build GRCh38
rs115812370 Chr.5: 134566333-134566333 on Build GRCh38
rs753055946 Chr.5: 134566406-134566406 on Build GRCh38
rs372140466 Chr.5: 134566273-134566273 on Build GRCh38
rs571104259 Chr.5: 134566280-134566280 on Build GRCh38
rs369345598 Chr.5: 134566408-134566408 on Build GRCh38
rs761382043 Chr.5: 134566362-134566362 on Build GRCh38
rs115315395 Chr.5: 134566249-134566249 on Build GRCh38
rs1028692168 Chr.5: 134566403-134566403 on Build GRCh38
rs562405170 Chr.5: 134566415-134566415 on Build GRCh38
rs763747988 Chr.5: 134566352-134566352 on Build GRCh38
rs779470752 Chr.5: 134566255-134566255 on Build GRCh38
rs200614486 Chr.5: 134566267-134566267 on Build GRCh38
rs748349072 Chr.5: 134566263-134566263 on Build GRCh38
rs762681690 Chr.5: 134566334-134566334 on Build GRCh38
rs141532868 Chr.5: 134566247-134566247 on Build GRCh38
rs957779421 Chr.5: 134566314-134566314 on Build GRCh38
rs751180082 Chr.5: 134566361-134566361 on Build GRCh38
rs906068716 Chr.5: 134566245-134566245 on Build GRCh38
rs142435802 Chr.5: 134566376-134566376 on Build GRCh38
rs550192691 Chr.5: 134566414-134566414 on Build GRCh38
rs1020255402 Chr.5: 134566358-134566358 on Build GRCh38
rs780644903 Chr.5: 134566252-134566252 on Build GRCh38
rs148155246 Chr.5: 134566244-134566244 on Build GRCh38
rs1002315432 Chr.5: 134566297-134566297 on Build GRCh38
rs895786070 Chr.5: 134566266-134566266 on Build GRCh38
rs755583646 Chr.5: 134566397-134566397 on Build GRCh38
rs372627663 Chr.5: 134566418-134566418 on Build GRCh38
rs976598787 Chr.5: 134566393-134566393 on Build GRCh38
rs1009290074 Chr.5: 134566347-134566347 on Build GRCh38
rs759525405 Chr.5: 134566310-134566310 on Build GRCh38
rs375321931 Chr.5: 134566298-134566298 on Build GRCh38
rs764992446 Chr.5: 134566332-134566332 on Build GRCh38
rs761984426 Chr.5: 134566401-134566401 on Build GRCh38

View Full Product Details