Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 27370537-27370774 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX17
Entrez Gene ID
9784
Gene Name
sorting nexin 17
Gene Aliases
-
UniGene
Hs.278569
Gene Chromosome Location
Chr.2: 27370496-27377533 on Build GRCh38
Transcripts
NM_001267059.1 NM_001267060.1 NM_001267061.1 NM_014748.3 NR_049782.1 NR_049783.1 NR_049784.1 NR_049785.1 NR_049786.1 NR_049787.1 NR_049788.1 XM_011533203.1 XM_017005405.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7316783 c.11C>T p.S4F Chr.2: 27370754-27370754 on Build GRCh38
4472702 c.17C>T p.P6L Chr.2: 27370760-27370760 on Build GRCh38
1690329 c.16C>T p.P6S Chr.2: 27370759-27370759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs557274905 Chr.2: 27370699-27370699 on Build GRCh38
rs751129915 Chr.2: 27370624-27370624 on Build GRCh38
rs775639243 Chr.2: 27370688-27370688 on Build GRCh38
rs968040114 Chr.2: 27370575-27370575 on Build GRCh38
rs781313455 Chr.2: 27370661-27370661 on Build GRCh38
rs1056663871 Chr.2: 27370650-27370650 on Build GRCh38
rs916340369 Chr.2: 27370631-27370631 on Build GRCh38
rs903889917 Chr.2: 27370676-27370676 on Build GRCh38
rs999783802 Chr.2: 27370590-27370590 on Build GRCh38
rs762210354 Chr.2: 27370720-27370720 on Build GRCh38
rs902724120 Chr.2: 27370735-27370735 on Build GRCh38
rs909207502 Chr.2: 27370681-27370681 on Build GRCh38
rs779801080 Chr.2: 27370644-27370644 on Build GRCh38
rs997365827 Chr.2: 27370694-27370694 on Build GRCh38
rs772195695 Chr.2: 27370683-27370683 on Build GRCh38
rs941666349 Chr.2: 27370619-27370619 on Build GRCh38
rs768687268 Chr.2: 27370700-27370700 on Build GRCh38
rs541521803 Chr.2: 27370721-27370722 on Build GRCh38
rs923049596 Chr.2: 27370595-27370595 on Build GRCh38
rs979758967 Chr.2: 27370578-27370578 on Build GRCh38
rs552210766 Chr.2: 27370586-27370586 on Build GRCh38
rs997620524 Chr.2: 27370715-27370715 on Build GRCh38
rs942054067 Chr.2: 27370684-27370684 on Build GRCh38
rs777167990 Chr.2: 27370714-27370714 on Build GRCh38
rs534682634 Chr.2: 27370640-27370640 on Build GRCh38
rs908953940 Chr.2: 27370736-27370736 on Build GRCh38
rs112575449 Chr.2: 27370581-27370581 on Build GRCh38
rs896625864 Chr.2: 27370654-27370654 on Build GRCh38
rs1009661540 Chr.2: 27370659-27370659 on Build GRCh38
rs926881965 Chr.2: 27370643-27370643 on Build GRCh38
rs957725987 Chr.2: 27370600-27370600 on Build GRCh38
rs918427091 Chr.2: 27370655-27370655 on Build GRCh38
rs144823348 Chr.2: 27370678-27370678 on Build GRCh38
rs975877212 Chr.2: 27370589-27370589 on Build GRCh38
rs983609673 Chr.2: 27370727-27370727 on Build GRCh38
rs751013608 Chr.2: 27370577-27370577 on Build GRCh38
rs11557230 Chr.2: 27370677-27370677 on Build GRCh38
rs752725431 Chr.2: 27370708-27370708 on Build GRCh38
rs748069063 Chr.2: 27370665-27370665 on Build GRCh38
rs1032010423 Chr.2: 27370597-27370597 on Build GRCh38
rs568708476 Chr.2: 27370559-27370559 on Build GRCh38
rs113964476 Chr.2: 27370716-27370716 on Build GRCh38
rs149434575 Chr.2: 27370601-27370601 on Build GRCh38
rs908878370 Chr.2: 27370617-27370617 on Build GRCh38
rs536149764 Chr.2: 27370692-27370692 on Build GRCh38

View Full Product Details