Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 150544349-150544608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SERP1
Entrez Gene ID
27230
Gene Name
stress-associated endoplasmic reticulum protein 1
Gene Aliases
RAMP4
UniGene
Hs.518326
Gene Chromosome Location
Chr.3: 150541993-150603177 on Build GRCh38
Transcripts
NM_014445.3 XM_011512698.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1617052 c.194G>A p.G65D Chr.3: 150544465-150544465 on Build GRCh38
3003030 c.171G>A p.Q57Q Chr.3: 150544488-150544488 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs762496300 Chr.3: 150544546-150544546 on Build GRCh38
rs751569122 Chr.3: 150544448-150544448 on Build GRCh38
rs957693409 Chr.3: 150544517-150544517 on Build GRCh38
rs369518471 Chr.3: 150544477-150544477 on Build GRCh38
rs759958005 Chr.3: 150544386-150544386 on Build GRCh38
rs369865086 Chr.3: 150544390-150544390 on Build GRCh38
rs779403834 Chr.3: 150544434-150544434 on Build GRCh38
rs377736024 Chr.3: 150544553-150544553 on Build GRCh38
rs527273162 Chr.3: 150544580-150544580 on Build GRCh38
rs778016891 Chr.3: 150544485-150544485 on Build GRCh38
rs749978281 Chr.3: 150544422-150544422 on Build GRCh38
rs1533816 Chr.3: 150544531-150544531 on Build GRCh38
rs764945316 Chr.3: 150544410-150544410 on Build GRCh38
rs780967478 Chr.3: 150544466-150544466 on Build GRCh38
rs561382047 Chr.3: 150544565-150544565 on Build GRCh38
rs749439404 Chr.3: 150544498-150544498 on Build GRCh38
rs774283378 Chr.3: 150544504-150544504 on Build GRCh38
rs772961214 Chr.3: 150544543-150544543 on Build GRCh38
rs893403326 Chr.3: 150544426-150544426 on Build GRCh38
rs761562880 Chr.3: 150544398-150544398 on Build GRCh38
rs1031981765 Chr.3: 150544508-150544508 on Build GRCh38
rs570559063 Chr.3: 150544514-150544514 on Build GRCh38
rs370298193 Chr.3: 150544425-150544425 on Build GRCh38
rs774962533 Chr.3: 150544371-150544371 on Build GRCh38
rs771108702 Chr.3: 150544502-150544502 on Build GRCh38
rs776031400 Chr.3: 150544530-150544530 on Build GRCh38
rs373546630 Chr.3: 150544513-150544513 on Build GRCh38
rs377408263 Chr.3: 150544527-150544527 on Build GRCh38
rs770893592 Chr.3: 150544370-150544370 on Build GRCh38
rs760411512 Chr.3: 150544374-150544375 on Build GRCh38
rs770023979 Chr.3: 150544479-150544479 on Build GRCh38
rs764536086 Chr.3: 150544533-150544533 on Build GRCh38
rs768053304 Chr.3: 150544387-150544387 on Build GRCh38
rs147209818 Chr.3: 150544464-150544464 on Build GRCh38

View Full Product Details