Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 15897787-15898058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
STRAP
Entrez Gene ID
11171
Gene Name
serine/threonine kinase receptor associated protein
Gene Aliases
MAWD, PT-WD, UNRIP
UniGene
Hs.743971
Gene Chromosome Location
Chr.12: 15882354-15903478 on Build GRCh38
Transcripts
NM_007178.3 XM_005253294.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5577096 c.724G>A p.G242S Chr.12: 15897967-15897967 on Build GRCh38
3458678 c.657C>T p.S219S Chr.12: 15897900-15897900 on Build GRCh38
3811598 c.657C>A p.S219S Chr.12: 15897900-15897900 on Build GRCh38
937637 c.750T>C p.Y250Y Chr.12: 15897993-15897993 on Build GRCh38
3458679 c.694C>T p.H232Y Chr.12: 15897937-15897937 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964213648 Chr.12: 15897830-15897830 on Build GRCh38
rs942338975 Chr.12: 15897888-15897888 on Build GRCh38
rs754719468 Chr.12: 15897974-15897974 on Build GRCh38
rs773312361 Chr.12: 15897870-15897870 on Build GRCh38
rs759466450 Chr.12: 15897966-15897966 on Build GRCh38
rs10772879 Chr.12: 15897874-15897874 on Build GRCh38
rs756674544 Chr.12: 15897944-15897947 on Build GRCh38
rs1005622113 Chr.12: 15897821-15897821 on Build GRCh38
rs778553680 Chr.12: 15897842-15897842 on Build GRCh38
rs765215889 Chr.12: 15897899-15897899 on Build GRCh38
rs867043357 Chr.12: 15897839-15897839 on Build GRCh38
rs372833344 Chr.12: 15897878-15897878 on Build GRCh38
rs185224819 Chr.12: 15897967-15897967 on Build GRCh38
rs778047951 Chr.12: 15897829-15897829 on Build GRCh38
rs145164927 Chr.12: 15897937-15897937 on Build GRCh38
rs531686762 Chr.12: 15898003-15898003 on Build GRCh38
rs752866252 Chr.12: 15897916-15897916 on Build GRCh38
rs778264792 Chr.12: 15897993-15897993 on Build GRCh38
rs759900698 Chr.12: 15897881-15897881 on Build GRCh38
rs772096190 Chr.12: 15897869-15897869 on Build GRCh38
rs752087716 Chr.12: 15897945-15897945 on Build GRCh38
rs764626491 Chr.12: 15897941-15897941 on Build GRCh38
rs987742946 Chr.12: 15898002-15898002 on Build GRCh38
rs748023580 Chr.12: 15897850-15897850 on Build GRCh38
rs760497317 Chr.12: 15897872-15897872 on Build GRCh38
rs577728776 Chr.12: 15897833-15897833 on Build GRCh38
rs767985267 Chr.12: 15897841-15897841 on Build GRCh38

View Full Product Details