Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44136256-44136517 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM63B
Entrez Gene ID
55362
Gene Name
transmembrane protein 63B
Gene Aliases
C6orf110
UniGene
Hs.414473
Gene Chromosome Location
Chr.6: 44126628-44155519 on Build GRCh38
Transcripts
NM_001318792.1 NM_018426.2 XM_005249211.3 XM_005249213.4 XM_005249214.4 XM_005249215.4 XM_005249217.1 XM_006715135.1 XM_017010999.1 XM_017011000.1 XM_017011001.1 XM_017011002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5781963 c.280G>T p.V94L Chr.6: 44136350-44136350 on Build GRCh38
1176462 c.322C>T p.R108C Chr.6: 44136392-44136392 on Build GRCh38
5764011 c.369+3G>T p.? Chr.6: 44136442-44136442 on Build GRCh38
7349524 c.287C>T p.S96L Chr.6: 44136357-44136357 on Build GRCh38
3352214 c.298G>A p.G100R Chr.6: 44136368-44136368 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753747915 Chr.6: 44136412-44136412 on Build GRCh38
rs746513634 Chr.6: 44136344-44136344 on Build GRCh38
rs773389164 Chr.6: 44136488-44136488 on Build GRCh38
rs767772279 Chr.6: 44136485-44136485 on Build GRCh38
rs1055977495 Chr.6: 44136456-44136456 on Build GRCh38
rs928890262 Chr.6: 44136361-44136361 on Build GRCh38
rs190666305 Chr.6: 44136455-44136455 on Build GRCh38
rs778862593 Chr.6: 44136416-44136416 on Build GRCh38
rs200329482 Chr.6: 44136324-44136324 on Build GRCh38
rs370393370 Chr.6: 44136297-44136297 on Build GRCh38
rs779838270 Chr.6: 44136308-44136308 on Build GRCh38
rs762212318 Chr.6: 44136312-44136312 on Build GRCh38
rs543923696 Chr.6: 44136452-44136452 on Build GRCh38
rs866230781 Chr.6: 44136405-44136405 on Build GRCh38
rs766375617 Chr.6: 44136393-44136393 on Build GRCh38
rs747884737 Chr.6: 44136429-44136429 on Build GRCh38
rs773108945 Chr.6: 44136450-44136451 on Build GRCh38
rs770752853 Chr.6: 44136469-44136474 on Build GRCh38
rs771791734 Chr.6: 44136333-44136333 on Build GRCh38
rs749153961 Chr.6: 44136309-44136309 on Build GRCh38
rs770391172 Chr.6: 44136345-44136345 on Build GRCh38
rs774989257 Chr.6: 44136290-44136290 on Build GRCh38
rs376470724 Chr.6: 44136353-44136353 on Build GRCh38
rs758191592 Chr.6: 44136451-44136451 on Build GRCh38
rs143570588 Chr.6: 44136415-44136415 on Build GRCh38
rs368623210 Chr.6: 44136329-44136329 on Build GRCh38
rs199751940 Chr.6: 44136352-44136352 on Build GRCh38
rs769151737 Chr.6: 44136461-44136461 on Build GRCh38
rs774651634 Chr.6: 44136367-44136367 on Build GRCh38
rs762039391 Chr.6: 44136368-44136368 on Build GRCh38
rs965908074 Chr.6: 44136288-44136288 on Build GRCh38
rs746984534 Chr.6: 44136451-44136451 on Build GRCh38
rs934631039 Chr.6: 44136365-44136365 on Build GRCh38
rs531946658 Chr.6: 44136362-44136362 on Build GRCh38
rs1047575130 Chr.6: 44136496-44136496 on Build GRCh38
rs767663908 Chr.6: 44136383-44136383 on Build GRCh38
rs756120817 Chr.6: 44136385-44136385 on Build GRCh38
rs756086085 Chr.6: 44136298-44136298 on Build GRCh38
rs146246234 Chr.6: 44136314-44136314 on Build GRCh38
rs771590105 Chr.6: 44136417-44136418 on Build GRCh38
rs746567184 Chr.6: 44136454-44136454 on Build GRCh38
rs372880804 Chr.6: 44136343-44136343 on Build GRCh38
rs201591306 Chr.6: 44136483-44136483 on Build GRCh38
rs772851514 Chr.6: 44136337-44136337 on Build GRCh38
rs750572327 Chr.6: 44136384-44136384 on Build GRCh38
rs995626519 Chr.6: 44136489-44136489 on Build GRCh38

View Full Product Details