Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72742323-72742584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMC1
Entrez Gene ID
117531
Gene Name
transmembrane channel like 1
Gene Aliases
DFNA36, DFNB11, DFNB7
UniGene
Hs.670211
Gene Chromosome Location
Chr.9: 72521801-72836351 on Build GRCh38
Transcripts
NM_138691.2 XM_017014256.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4400366 c.480T>A p.F160L Chr.9: 72742470-72742470 on Build GRCh38
1132457 c.475G>A p.D159N Chr.9: 72742465-72742465 on Build GRCh38
6802760 c.473G>A p.R158H Chr.9: 72742463-72742463 on Build GRCh38
5024250 c.535+6T>C p.? Chr.9: 72742531-72742531 on Build GRCh38
6115640 c.496G>T p.A166S Chr.9: 72742486-72742486 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777607381 Chr.9: 72742537-72742537 on Build GRCh38
rs371485372 Chr.9: 72742451-72742451 on Build GRCh38
rs1010005378 Chr.9: 72742412-72742413 on Build GRCh38
rs762864983 Chr.9: 72742522-72742523 on Build GRCh38
rs757513819 Chr.9: 72742554-72742554 on Build GRCh38
rs753562476 Chr.9: 72742540-72742540 on Build GRCh38
rs767019298 Chr.9: 72742497-72742497 on Build GRCh38
rs200841002 Chr.9: 72742462-72742462 on Build GRCh38
rs375984349 Chr.9: 72742473-72742473 on Build GRCh38
rs765014770 Chr.9: 72742396-72742396 on Build GRCh38
rs563498098 Chr.9: 72742397-72742397 on Build GRCh38
rs373823051 Chr.9: 72742545-72742545 on Build GRCh38
rs899519982 Chr.9: 72742402-72742402 on Build GRCh38
rs756028904 Chr.9: 72742519-72742519 on Build GRCh38
rs765971881 Chr.9: 72742406-72742406 on Build GRCh38
rs765885670 Chr.9: 72742487-72742487 on Build GRCh38
rs547462602 Chr.9: 72742395-72742395 on Build GRCh38
rs762736726 Chr.9: 72742483-72742483 on Build GRCh38
rs762674080 Chr.9: 72742404-72742404 on Build GRCh38
rs13302809 Chr.9: 72742413-72742413 on Build GRCh38
rs921146046 Chr.9: 72742353-72742353 on Build GRCh38
rs1051044239 Chr.9: 72742376-72742376 on Build GRCh38
rs796412699 Chr.9: 72742492-72742492 on Build GRCh38
rs749566965 Chr.9: 72742422-72742422 on Build GRCh38
rs148340276 Chr.9: 72742463-72742463 on Build GRCh38
rs551241375 Chr.9: 72742364-72742364 on Build GRCh38
rs752678337 Chr.9: 72742499-72742499 on Build GRCh38
rs775383060 Chr.9: 72742432-72742432 on Build GRCh38
rs772364280 Chr.9: 72742528-72742528 on Build GRCh38
rs886042321 Chr.9: 72742524-72742524 on Build GRCh38
rs777517407 Chr.9: 72742418-72742418 on Build GRCh38
rs376848757 Chr.9: 72742409-72742409 on Build GRCh38
rs527751053 Chr.9: 72742392-72742392 on Build GRCh38
rs117399674 Chr.9: 72742425-72742425 on Build GRCh38
rs761235546 Chr.9: 72742458-72742458 on Build GRCh38
rs745876096 Chr.9: 72742433-72742433 on Build GRCh38
rs751031180 Chr.9: 72742407-72742407 on Build GRCh38
rs747645067 Chr.9: 72742520-72742520 on Build GRCh38
rs145102585 Chr.9: 72742450-72742450 on Build GRCh38
rs370032879 Chr.9: 72742488-72742488 on Build GRCh38
rs758924923 Chr.9: 72742494-72742494 on Build GRCh38
rs755941190 Chr.9: 72742417-72742417 on Build GRCh38
rs199771068 Chr.9: 72742414-72742414 on Build GRCh38
rs771053186 Chr.9: 72742424-72742424 on Build GRCh38
rs539228804 Chr.9: 72742452-72742452 on Build GRCh38
rs565049075 Chr.9: 72742389-72742389 on Build GRCh38

View Full Product Details