Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 72210608-72210853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GDA
Entrez Gene ID
9615
Gene Name
guanine deaminase
Gene Aliases
CYPIN, GUANASE, NEDASIN
UniGene
Hs.494163
Gene Chromosome Location
Chr.9: 72114595-72256045 on Build GRCh38
Transcripts
NM_001242505.2 NM_001242506.2 NM_001242507.2 NM_004293.4 XM_005252317.1 XM_011519213.1 XM_011519214.1 XM_011519215.1 XM_011519216.1 XM_011519217.1 XM_011519218.2 XM_011519219.1 XM_011519220.1 XM_011519221.1 XM_011519222.1 XM_011519223.2 XM_017015333.1 XM_017015334.1 XM_017015335.1 XM_017015336.1 XM_017015337.1 XM_017015338.1 XM_017015339.1 XM_017015340.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2780019 c.463G>A p.D155N Chr.9: 72210765-72210765 on Build GRCh38
3358365 c.447A>C p.S149S Chr.9: 72210749-72210749 on Build GRCh38
3926920 c.411C>T p.T137T Chr.9: 72210713-72210713 on Build GRCh38
3926921 c.411C>T p.T137T Chr.9: 72210713-72210713 on Build GRCh38
5411215 c.409A>G p.T137A Chr.9: 72210711-72210711 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766003586 Chr.9: 72210757-72210757 on Build GRCh38
rs745944565 Chr.9: 72210798-72210798 on Build GRCh38
rs772769043 Chr.9: 72210742-72210742 on Build GRCh38
rs148805997 Chr.9: 72210698-72210698 on Build GRCh38
rs749072901 Chr.9: 72210790-72210790 on Build GRCh38
rs777667197 Chr.9: 72210768-72210768 on Build GRCh38
rs201591902 Chr.9: 72210662-72210662 on Build GRCh38
rs752635142 Chr.9: 72210765-72210765 on Build GRCh38
rs747509974 Chr.9: 72210805-72210805 on Build GRCh38
rs759240376 Chr.9: 72210663-72210663 on Build GRCh38
rs781104187 Chr.9: 72210649-72210649 on Build GRCh38
rs775995372 Chr.9: 72210712-72210712 on Build GRCh38
rs767369489 Chr.9: 72210764-72210764 on Build GRCh38
rs777912068 Chr.9: 72210655-72210655 on Build GRCh38
rs751138545 Chr.9: 72210758-72210758 on Build GRCh38
rs745926375 Chr.9: 72210689-72210689 on Build GRCh38
rs375150689 Chr.9: 72210665-72210665 on Build GRCh38
rs11143170 Chr.9: 72210791-72210791 on Build GRCh38
rs764471126 Chr.9: 72210732-72210732 on Build GRCh38
rs772217332 Chr.9: 72210801-72210801 on Build GRCh38
rs754418920 Chr.9: 72210763-72210763 on Build GRCh38
rs990561052 Chr.9: 72210770-72210770 on Build GRCh38
rs905885932 Chr.9: 72210752-72210752 on Build GRCh38
rs530453363 Chr.9: 72210664-72210664 on Build GRCh38
rs755884857 Chr.9: 72210656-72210656 on Build GRCh38
rs779271384 Chr.9: 72210792-72210792 on Build GRCh38
rs760960652 Chr.9: 72210721-72210721 on Build GRCh38
rs372575836 Chr.9: 72210772-72210772 on Build GRCh38
rs775881671 Chr.9: 72210804-72210804 on Build GRCh38
rs762633583 Chr.9: 72210746-72210746 on Build GRCh38
rs769012935 Chr.9: 72210826-72210826 on Build GRCh38

View Full Product Details