Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219505565-219505808 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GMPPA
Entrez Gene ID
29926
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
AAMR
UniGene
Hs.27059
Gene Chromosome Location
Chr.2: 219498865-219506996 on Build GRCh38
Transcripts
NM_013335.3 NM_205847.2 XM_005246483.3 XM_005246486.3 XM_011511032.2 XM_017003906.1 XM_017003907.1 XM_017003908.1 XM_017003909.1 XM_017003910.1 XM_017003911.1 XM_017003912.1 XM_017003913.1 XM_017003914.1 XM_017003915.1 XM_017003916.1 XM_017003917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6672041 c.867C>G p.I289M Chr.2: 219505728-219505728 on Build GRCh38
4091723 c.872C>T p.P291L Chr.2: 219505733-219505733 on Build GRCh38
5657358 c.858T>G p.N286K Chr.2: 219505719-219505719 on Build GRCh38
1208380 c.863A>G p.Y288C Chr.2: 219505724-219505724 on Build GRCh38
1016824 c.873G>A p.P291P Chr.2: 219505734-219505734 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs953842015 Chr.2: 219505612-219505612 on Build GRCh38
rs762029038 Chr.2: 219505626-219505626 on Build GRCh38
rs776373574 Chr.2: 219505610-219505610 on Build GRCh38
rs961563489 Chr.2: 219505665-219505665 on Build GRCh38
rs746804961 Chr.2: 219505591-219505591 on Build GRCh38
rs550099366 Chr.2: 219505587-219505587 on Build GRCh38
rs761066072 Chr.2: 219505650-219505650 on Build GRCh38
rs371527019 Chr.2: 219505737-219505737 on Build GRCh38
rs769745264 Chr.2: 219505734-219505734 on Build GRCh38
rs766774052 Chr.2: 219505657-219505657 on Build GRCh38
rs776643066 Chr.2: 219505699-219505699 on Build GRCh38
rs372564894 Chr.2: 219505606-219505606 on Build GRCh38
rs903900192 Chr.2: 219505772-219505772 on Build GRCh38
rs1017038834 Chr.2: 219505653-219505653 on Build GRCh38
rs754262824 Chr.2: 219505662-219505662 on Build GRCh38
rs145760617 Chr.2: 219505719-219505719 on Build GRCh38
rs151043830 Chr.2: 219505738-219505738 on Build GRCh38
rs777036634 Chr.2: 219505759-219505759 on Build GRCh38
rs955950736 Chr.2: 219505586-219505586 on Build GRCh38
rs770772480 Chr.2: 219505608-219505608 on Build GRCh38
rs1005620276 Chr.2: 219505640-219505640 on Build GRCh38
rs751576633 Chr.2: 219505611-219505611 on Build GRCh38
rs61304293 Chr.2: 219505692-219505692 on Build GRCh38
rs773633201 Chr.2: 219505632-219505632 on Build GRCh38
rs575126365 Chr.2: 219505781-219505781 on Build GRCh38
rs554955899 Chr.2: 219505768-219505768 on Build GRCh38
rs762393620 Chr.2: 219505785-219505785 on Build GRCh38
rs752221989 Chr.2: 219505673-219505673 on Build GRCh38
rs746676212 Chr.2: 219505705-219505705 on Build GRCh38
rs761012765 Chr.2: 219505754-219505754 on Build GRCh38
rs972393344 Chr.2: 219505694-219505694 on Build GRCh38
rs768577936 Chr.2: 219505617-219505617 on Build GRCh38
rs745743360 Chr.2: 219505733-219505733 on Build GRCh38
rs755535192 Chr.2: 219505672-219505672 on Build GRCh38
rs781071244 Chr.2: 219505725-219505725 on Build GRCh38
rs773504203 Chr.2: 219505750-219505750 on Build GRCh38
rs772333659 Chr.2: 219505742-219505742 on Build GRCh38
rs771155229 Chr.2: 219505755-219505755 on Build GRCh38
rs759858230 Chr.2: 219505765-219505765 on Build GRCh38
rs569722184 Chr.2: 219505624-219505624 on Build GRCh38
rs757972361 Chr.2: 219505677-219505677 on Build GRCh38

View Full Product Details