Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 8313386-8313659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ARHGEF15
Entrez Gene ID
22899
Gene Name
Rho guanine nucleotide exchange factor 15
Gene Aliases
ARGEF15, E5, Ephexin5, Vsm-RhoGEF
UniGene
Hs.443109
Gene Chromosome Location
Chr.17: 8310238-8322516 on Build GRCh38
Transcripts
NM_025014.1 NM_173728.3 XM_011523734.1 XM_011523735.1 XM_011523736.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3821099 c.970C>G p.P324A Chr.17: 8313536-8313536 on Build GRCh38
4973365 c.949G>A p.E317K Chr.17: 8313515-8313515 on Build GRCh38
109744 c.936G>A p.G312G Chr.17: 8313502-8313502 on Build GRCh38
3932793 c.948T>C p.D316D Chr.17: 8313514-8313514 on Build GRCh38
344622 c.936G>T p.G312G Chr.17: 8313502-8313502 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs758937731 Chr.17: 8313593-8313593 on Build GRCh38
rs531197696 Chr.17: 8313504-8313504 on Build GRCh38
rs944918976 Chr.17: 8313480-8313480 on Build GRCh38
rs879117782 Chr.17: 8313430-8313430 on Build GRCh38
rs376805138 Chr.17: 8313501-8313501 on Build GRCh38
rs780440591 Chr.17: 8313513-8313513 on Build GRCh38
rs954004122 Chr.17: 8313616-8313616 on Build GRCh38
rs2302866 Chr.17: 8313440-8313440 on Build GRCh38
rs939849332 Chr.17: 8313622-8313622 on Build GRCh38
rs762656286 Chr.17: 8313458-8313458 on Build GRCh38
rs774020684 Chr.17: 8313568-8313568 on Build GRCh38
rs759067562 Chr.17: 8313474-8313474 on Build GRCh38
rs750671470 Chr.17: 8313503-8313503 on Build GRCh38
rs756469375 Chr.17: 8313524-8313524 on Build GRCh38
rs773497449 Chr.17: 8313560-8313560 on Build GRCh38
rs2302867 Chr.17: 8313419-8313419 on Build GRCh38
rs780532902 Chr.17: 8313457-8313457 on Build GRCh38
rs563543358 Chr.17: 8313482-8313482 on Build GRCh38
rs755121072 Chr.17: 8313518-8313518 on Build GRCh38
rs893447741 Chr.17: 8313481-8313481 on Build GRCh38
rs974756472 Chr.17: 8313486-8313486 on Build GRCh38
rs985326074 Chr.17: 8313624-8313624 on Build GRCh38
rs777540804 Chr.17: 8313474-8313475 on Build GRCh38
rs747736264 Chr.17: 8313465-8313466 on Build GRCh38
rs747039155 Chr.17: 8313515-8313515 on Build GRCh38
rs775276662 Chr.17: 8313604-8313604 on Build GRCh38
rs373347356 Chr.17: 8313459-8313459 on Build GRCh38
rs773789933 Chr.17: 8313473-8313473 on Build GRCh38
rs930752469 Chr.17: 8313582-8313582 on Build GRCh38
rs755702728 Chr.17: 8313466-8313466 on Build GRCh38
rs755842800 Chr.17: 8313500-8313500 on Build GRCh38
rs748744226 Chr.17: 8313540-8313540 on Build GRCh38
rs1049136398 Chr.17: 8313588-8313588 on Build GRCh38
rs767318015 Chr.17: 8313485-8313485 on Build GRCh38
rs138318077 Chr.17: 8313502-8313502 on Build GRCh38
rs372368712 Chr.17: 8313475-8313476 on Build GRCh38
rs770046438 Chr.17: 8313550-8313550 on Build GRCh38
rs966449328 Chr.17: 8313562-8313562 on Build GRCh38
rs75233652 Chr.17: 8313601-8313601 on Build GRCh38
rs760732807 Chr.17: 8313606-8313606 on Build GRCh38
rs749663183 Chr.17: 8313564-8313564 on Build GRCh38
rs112237413 Chr.17: 8313526-8313526 on Build GRCh38
rs752719110 Chr.17: 8313489-8313489 on Build GRCh38
rs770521016 Chr.17: 8313567-8313567 on Build GRCh38

View Full Product Details