Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 181474941-181475176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA4
Entrez Gene ID
3676
Gene Name
integrin subunit alpha 4
Gene Aliases
CD49D, IA4
UniGene
Hs.440955
Gene Chromosome Location
Chr.2: 181456892-181538928 on Build GRCh38
Transcripts
NM_000885.5 NM_001316312.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6493425 c.378G>C p.W126C Chr.2: 181475018-181475018 on Build GRCh38
6089148 c.425T>A p.V142E Chr.2: 181475065-181475065 on Build GRCh38
3425828 c.430T>G p.C144G Chr.2: 181475162-181475162 on Build GRCh38
4825125 c.361G>C p.E121Q Chr.2: 181475001-181475001 on Build GRCh38
7372558 c.350C>A p.T117N Chr.2: 181474990-181474990 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201604237 Chr.2: 181474977-181474977 on Build GRCh38
rs558463198 Chr.2: 181475080-181475080 on Build GRCh38
rs775276826 Chr.2: 181475110-181475110 on Build GRCh38
rs201805626 Chr.2: 181475074-181475074 on Build GRCh38
rs766805617 Chr.2: 181475130-181475130 on Build GRCh38
rs746714668 Chr.2: 181475112-181475112 on Build GRCh38
rs756147388 Chr.2: 181475091-181475091 on Build GRCh38
rs199774682 Chr.2: 181474979-181474979 on Build GRCh38
rs766895491 Chr.2: 181475076-181475076 on Build GRCh38
rs200403982 Chr.2: 181475098-181475098 on Build GRCh38
rs774236136 Chr.2: 181475007-181475007 on Build GRCh38
rs201754311 Chr.2: 181475123-181475123 on Build GRCh38
rs202180944 Chr.2: 181475096-181475096 on Build GRCh38
rs200029223 Chr.2: 181475084-181475084 on Build GRCh38
rs763727240 Chr.2: 181475039-181475039 on Build GRCh38
rs200989622 Chr.2: 181475086-181475086 on Build GRCh38
rs200580619 Chr.2: 181474994-181474994 on Build GRCh38
rs745596932 Chr.2: 181475107-181475107 on Build GRCh38
rs760089206 Chr.2: 181475030-181475030 on Build GRCh38
rs904544405 Chr.2: 181475047-181475047 on Build GRCh38
rs1009330577 Chr.2: 181475004-181475004 on Build GRCh38
rs767670080 Chr.2: 181475085-181475085 on Build GRCh38
rs949026133 Chr.2: 181475124-181475124 on Build GRCh38
rs778995676 Chr.2: 181475103-181475103 on Build GRCh38
rs199686726 Chr.2: 181475142-181475142 on Build GRCh38
rs968533783 Chr.2: 181475095-181475095 on Build GRCh38
rs1007677551 Chr.2: 181474995-181474995 on Build GRCh38
rs267599105 Chr.2: 181475056-181475056 on Build GRCh38
rs745686958 Chr.2: 181475024-181475024 on Build GRCh38
rs772047310 Chr.2: 181475108-181475108 on Build GRCh38
rs372164526 Chr.2: 181475029-181475029 on Build GRCh38
rs752781224 Chr.2: 181475141-181475141 on Build GRCh38
rs774853787 Chr.2: 181475137-181475137 on Build GRCh38
rs201346236 Chr.2: 181475064-181475064 on Build GRCh38
rs537730254 Chr.2: 181475094-181475094 on Build GRCh38
rs1017590073 Chr.2: 181475018-181475018 on Build GRCh38
rs866781844 Chr.2: 181475042-181475042 on Build GRCh38
rs199901297 Chr.2: 181475063-181475063 on Build GRCh38
rs201407824 Chr.2: 181475006-181475006 on Build GRCh38
rs201075106 Chr.2: 181475113-181475113 on Build GRCh38
rs180755677 Chr.2: 181475116-181475116 on Build GRCh38
rs749317117 Chr.2: 181474984-181474984 on Build GRCh38
rs200238112 Chr.2: 181475122-181475122 on Build GRCh38

View Full Product Details