Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 108884026-108884285 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C7orf66
Entrez Gene ID
154907
Gene Name
chromosome 7 open reading frame 66
Gene Aliases
-
UniGene
Hs.276325
Gene Chromosome Location
Chr.7: 108883975-108884587 on Build GRCh38
Transcripts
NM_001024607.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6616662 c.234C>T p.Y78Y Chr.7: 108884121-108884121 on Build GRCh38
7096744 c.250G>A p.E84K Chr.7: 108884105-108884105 on Build GRCh38
4649043 c.209A>G p.Y70C Chr.7: 108884146-108884146 on Build GRCh38
4654165 c.265C>T p.Q89* Chr.7: 108884090-108884090 on Build GRCh38
4668016 c.316G>A p.A106T Chr.7: 108884039-108884039 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763850896 Chr.7: 108884195-108884195 on Build GRCh38
rs911070475 Chr.7: 108884114-108884114 on Build GRCh38
rs1046769542 Chr.7: 108884179-108884179 on Build GRCh38
rs781619382 Chr.7: 108884143-108884143 on Build GRCh38
rs148085892 Chr.7: 108884188-108884188 on Build GRCh38
rs759073131 Chr.7: 108884172-108884172 on Build GRCh38
rs879369674 Chr.7: 108884183-108884183 on Build GRCh38
rs745902325 Chr.7: 108884113-108884113 on Build GRCh38
rs752425624 Chr.7: 108884068-108884068 on Build GRCh38
rs929740675 Chr.7: 108884176-108884176 on Build GRCh38
rs922377014 Chr.7: 108884155-108884155 on Build GRCh38
rs750366015 Chr.7: 108884218-108884218 on Build GRCh38
rs942275181 Chr.7: 108884217-108884217 on Build GRCh38
rs367934561 Chr.7: 108884174-108884174 on Build GRCh38
rs769225310 Chr.7: 108884239-108884240 on Build GRCh38
rs1012859561 Chr.7: 108884242-108884242 on Build GRCh38
rs767406611 Chr.7: 108884214-108884214 on Build GRCh38
rs756129528 Chr.7: 108884234-108884234 on Build GRCh38
rs768981934 Chr.7: 108884148-108884148 on Build GRCh38
rs370890601 Chr.7: 108884080-108884080 on Build GRCh38
rs762498181 Chr.7: 108884194-108884194 on Build GRCh38
rs905197566 Chr.7: 108884187-108884187 on Build GRCh38
rs376651211 Chr.7: 108884144-108884144 on Build GRCh38
rs1037934639 Chr.7: 108884228-108884228 on Build GRCh38
rs532609676 Chr.7: 108884166-108884166 on Build GRCh38
rs375280615 Chr.7: 108884116-108884116 on Build GRCh38
rs139475645 Chr.7: 108884252-108884252 on Build GRCh38
rs757102533 Chr.7: 108884203-108884203 on Build GRCh38
rs978536662 Chr.7: 108884085-108884085 on Build GRCh38
rs989328733 Chr.7: 108884099-108884099 on Build GRCh38
rs751426871 Chr.7: 108884073-108884073 on Build GRCh38
rs143724624 Chr.7: 108884069-108884069 on Build GRCh38
rs772434368 Chr.7: 108884170-108884170 on Build GRCh38
rs781243164 Chr.7: 108884094-108884094 on Build GRCh38
rs751292803 Chr.7: 108884200-108884200 on Build GRCh38
rs764840851 Chr.7: 108884173-108884173 on Build GRCh38
rs748583087 Chr.7: 108884167-108884167 on Build GRCh38
rs756221351 Chr.7: 108884135-108884135 on Build GRCh38
rs942549194 Chr.7: 108884129-108884129 on Build GRCh38
rs1030920110 Chr.7: 108884086-108884086 on Build GRCh38
rs888081116 Chr.7: 108884201-108884201 on Build GRCh38
rs909697054 Chr.7: 108884152-108884152 on Build GRCh38
rs777440161 Chr.7: 108884072-108884072 on Build GRCh38
rs749698638 Chr.7: 108884124-108884125 on Build GRCh38
rs552878028 Chr.7: 108884062-108884062 on Build GRCh38
rs199705089 Chr.7: 108884090-108884090 on Build GRCh38

View Full Product Details