Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 94603182-94603431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FBP1
Entrez Gene ID
2203
Gene Name
fructose-bisphosphatase 1
Gene Aliases
FBP
UniGene
Hs.494496
Gene Chromosome Location
Chr.9: 94603133-94640257 on Build GRCh38
Transcripts
NM_000507.3 NM_001127628.1 XM_006717005.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4514687 c.984C>T p.F328F Chr.9: 94603414-94603414 on Build GRCh38
4187967 c.989_992delAGGT p.K330fs*>8 Chr.9: 94603406-94603409 on Build GRCh38
6502739 c.1007C>T p.S336F Chr.9: 94603391-94603391 on Build GRCh38
7401031 c.974T>A p.V325E Chr.9: 94603424-94603424 on Build GRCh38
3780211 c.976C>A p.L326I Chr.9: 94603422-94603422 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774919384 Chr.9: 94603208-94603208 on Build GRCh38
rs778406463 Chr.9: 94603333-94603333 on Build GRCh38
rs911113887 Chr.9: 94603212-94603212 on Build GRCh38
rs1062483 Chr.9: 94603364-94603364 on Build GRCh38
rs1042189 Chr.9: 94603255-94603255 on Build GRCh38
rs752198321 Chr.9: 94603349-94603349 on Build GRCh38
rs564777022 Chr.9: 94603273-94603273 on Build GRCh38
rs374715941 Chr.9: 94603377-94603377 on Build GRCh38
rs371737592 Chr.9: 94603372-94603372 on Build GRCh38
rs190431674 Chr.9: 94603229-94603229 on Build GRCh38
rs1035026871 Chr.9: 94603359-94603359 on Build GRCh38
rs930068556 Chr.9: 94603269-94603269 on Build GRCh38
rs528042884 Chr.9: 94603353-94603353 on Build GRCh38
rs891920976 Chr.9: 94603238-94603238 on Build GRCh38
rs907367042 Chr.9: 94603284-94603284 on Build GRCh38
rs768728743 Chr.9: 94603400-94603400 on Build GRCh38
rs1045449646 Chr.9: 94603375-94603375 on Build GRCh38
rs937244719 Chr.9: 94603394-94603394 on Build GRCh38
rs2274689 Chr.9: 94603260-94603260 on Build GRCh38
rs767071343 Chr.9: 94603340-94603340 on Build GRCh38
rs1054322109 Chr.9: 94603403-94603403 on Build GRCh38
rs1016604737 Chr.9: 94603390-94603390 on Build GRCh38
rs936029299 Chr.9: 94603223-94603223 on Build GRCh38
rs9695 Chr.9: 94603360-94603360 on Build GRCh38
rs1001686413 Chr.9: 94603285-94603285 on Build GRCh38
rs775783820 Chr.9: 94603385-94603385 on Build GRCh38
rs757595481 Chr.9: 94603365-94603365 on Build GRCh38
rs747059512 Chr.9: 94603397-94603397 on Build GRCh38
rs779355517 Chr.9: 94603369-94603369 on Build GRCh38
rs756684836 Chr.9: 94603350-94603350 on Build GRCh38
rs982857525 Chr.9: 94603281-94603281 on Build GRCh38

View Full Product Details