Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 40369963-40370202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PLD3
Entrez Gene ID
23646
Gene Name
phospholipase D family member 3
Gene Aliases
AD19, HU-K4, HUK4
UniGene
Hs.257008
Gene Chromosome Location
Chr.19: 40348395-40378490 on Build GRCh38
Transcripts
NM_001031696.3 NM_001291311.1 NM_012268.3 XM_005258704.1 XM_005258707.4 XM_005258708.3 XM_005258709.4 XM_005258710.4 XM_006723122.1 XM_011526692.1 XM_011526693.1 XM_017026546.1 XM_017026547.1 XM_017026548.1 XM_017026549.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6859648 c.391G>A p.G131S Chr.19: 40370028-40370028 on Build GRCh38
4502180 c.447C>T p.T149T Chr.19: 40370165-40370165 on Build GRCh38
6252241 c.463G>T p.D155Y Chr.19: 40370181-40370181 on Build GRCh38
3362856 c.580A>T p.K194* Chr.19: 40370139-40370139 on Build GRCh38
4498079 c.352C>T p.P118S Chr.19: 40369989-40369989 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775366093 Chr.19: 40370089-40370089 on Build GRCh38
rs745451549 Chr.19: 40370154-40370154 on Build GRCh38
rs764154244 Chr.19: 40370097-40370097 on Build GRCh38
rs773927974 Chr.19: 40370074-40370074 on Build GRCh38
rs748777623 Chr.19: 40370136-40370136 on Build GRCh38
rs779300090 Chr.19: 40369985-40369985 on Build GRCh38
rs750676918 Chr.19: 40370123-40370123 on Build GRCh38
rs565771988 Chr.19: 40370009-40370009 on Build GRCh38
rs780226569 Chr.19: 40370129-40370129 on Build GRCh38
rs181424414 Chr.19: 40370091-40370091 on Build GRCh38
rs771242533 Chr.19: 40370149-40370149 on Build GRCh38
rs931609223 Chr.19: 40370007-40370007 on Build GRCh38
rs759348378 Chr.19: 40370082-40370082 on Build GRCh38
rs747405880 Chr.19: 40370130-40370130 on Build GRCh38
rs780604999 Chr.19: 40370002-40370002 on Build GRCh38
rs770471881 Chr.19: 40370140-40370140 on Build GRCh38
rs775104213 Chr.19: 40370180-40370180 on Build GRCh38
rs757461460 Chr.19: 40370111-40370111 on Build GRCh38
rs777000690 Chr.19: 40370153-40370153 on Build GRCh38
rs1054134909 Chr.19: 40370044-40370044 on Build GRCh38
rs770527017 Chr.19: 40370073-40370073 on Build GRCh38
rs368065175 Chr.19: 40370003-40370003 on Build GRCh38
rs748828564 Chr.19: 40370071-40370071 on Build GRCh38
rs758727867 Chr.19: 40369991-40369991 on Build GRCh38
rs757143126 Chr.19: 40370044-40370045 on Build GRCh38
rs767285336 Chr.19: 40370087-40370087 on Build GRCh38
rs148056303 Chr.19: 40370122-40370122 on Build GRCh38
rs781526171 Chr.19: 40370047-40370047 on Build GRCh38
rs755438378 Chr.19: 40370133-40370133 on Build GRCh38
rs746220124 Chr.19: 40369988-40369988 on Build GRCh38
rs537389538 Chr.19: 40370098-40370098 on Build GRCh38
rs866850284 Chr.19: 40369995-40369995 on Build GRCh38
rs771756854 Chr.19: 40370177-40370177 on Build GRCh38
rs772643756 Chr.19: 40370066-40370066 on Build GRCh38
rs758774392 Chr.19: 40370126-40370126 on Build GRCh38
rs759940523 Chr.19: 40370166-40370166 on Build GRCh38
rs371987481 Chr.19: 40370027-40370027 on Build GRCh38
rs369743081 Chr.19: 40370026-40370026 on Build GRCh38
rs548202140 Chr.19: 40370045-40370045 on Build GRCh38
rs1053696523 Chr.19: 40370147-40370147 on Build GRCh38
rs781683397 Chr.19: 40370135-40370135 on Build GRCh38
rs774050277 Chr.19: 40370148-40370148 on Build GRCh38

View Full Product Details