Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 40818071-40818302 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNQ4
Entrez Gene ID
9132
Gene Name
potassium voltage-gated channel subfamily Q member 4
Gene Aliases
DFNA2, DFNA2A, KV7.4
UniGene
Hs.473058
Gene Chromosome Location
Chr.1: 40784012-40840457 on Build GRCh38
Transcripts
NM_004700.3 NM_172163.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
909496 c.430G>A p.G144S Chr.1: 40818188-40818188 on Build GRCh38
6311101 c.464G>T p.G155V Chr.1: 40818222-40818222 on Build GRCh38
133930 c.463G>A p.G155R Chr.1: 40818221-40818221 on Build GRCh38
5928098 c.518C>T p.P173L Chr.1: 40818276-40818276 on Build GRCh38
6691727 c.503G>A p.R168H Chr.1: 40818261-40818261 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745530561 Chr.1: 40818143-40818143 on Build GRCh38
rs762957513 Chr.1: 40818178-40818178 on Build GRCh38
rs200053059 Chr.1: 40818177-40818177 on Build GRCh38
rs759714910 Chr.1: 40818199-40818199 on Build GRCh38
rs727504635 Chr.1: 40818217-40818217 on Build GRCh38
rs766465736 Chr.1: 40818180-40818180 on Build GRCh38
rs1055199549 Chr.1: 40818099-40818099 on Build GRCh38
rs145129529 Chr.1: 40818223-40818223 on Build GRCh38
rs201982211 Chr.1: 40818152-40818152 on Build GRCh38
rs568067536 Chr.1: 40818155-40818155 on Build GRCh38
rs772605872 Chr.1: 40818254-40818259 on Build GRCh38
rs755979378 Chr.1: 40818203-40818203 on Build GRCh38
rs745510596 Chr.1: 40818283-40818283 on Build GRCh38
rs903942176 Chr.1: 40818169-40818169 on Build GRCh38
rs267598596 Chr.1: 40818164-40818164 on Build GRCh38
rs778668591 Chr.1: 40818141-40818141 on Build GRCh38
rs770347285 Chr.1: 40818161-40818161 on Build GRCh38
rs753849216 Chr.1: 40818228-40818228 on Build GRCh38
rs756787790 Chr.1: 40818230-40818230 on Build GRCh38
rs769043523 Chr.1: 40818149-40818149 on Build GRCh38
rs752503566 Chr.1: 40818202-40818202 on Build GRCh38
rs979744772 Chr.1: 40818259-40818259 on Build GRCh38
rs527870834 Chr.1: 40818115-40818115 on Build GRCh38
rs767337917 Chr.1: 40818200-40818200 on Build GRCh38
rs144434662 Chr.1: 40818196-40818196 on Build GRCh38
rs748748123 Chr.1: 40818114-40818114 on Build GRCh38
rs778605247 Chr.1: 40818267-40818267 on Build GRCh38

View Full Product Details