Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 59238413-59238644 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PHYHIPL
Entrez Gene ID
84457
Gene Name
phytanoyl-CoA 2-hydroxylase interacting protein like
Gene Aliases
-
UniGene
Hs.499704
Gene Chromosome Location
Chr.10: 59174811-59247774 on Build GRCh38
Transcripts
NM_001143774.1 NM_032439.3 XM_011540275.2 XM_011540276.2 XM_017016782.1 XM_017016783.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6745714 c.479-10delT p.? Chr.10: 59238578-59238578 on Build GRCh38
5492657 c.401-10_401-9insT p.? Chr.10: 59238578-59238579 on Build GRCh38
1165397 c.515A>T p.K172M Chr.10: 59238624-59238624 on Build GRCh38
6129758 c.532G>A p.G178R Chr.10: 59238641-59238641 on Build GRCh38
360732 c.535C>A p.R179S Chr.10: 59238644-59238644 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760460247 Chr.10: 59238604-59238604 on Build GRCh38
rs768534752 Chr.10: 59238613-59238613 on Build GRCh38
rs757720914 Chr.10: 59238616-59238616 on Build GRCh38
rs1056665349 Chr.10: 59238444-59238444 on Build GRCh38
rs747332267 Chr.10: 59238571-59238571 on Build GRCh38
rs572823217 Chr.10: 59238539-59238539 on Build GRCh38
rs3758565 Chr.10: 59238492-59238492 on Build GRCh38
rs769830481 Chr.10: 59238595-59238595 on Build GRCh38
rs913958148 Chr.10: 59238459-59238459 on Build GRCh38
rs778021003 Chr.10: 59238596-59238596 on Build GRCh38
rs191931300 Chr.10: 59238577-59238577 on Build GRCh38
rs376768188 Chr.10: 59238599-59238599 on Build GRCh38
rs73296198 Chr.10: 59238524-59238524 on Build GRCh38
rs201329709 Chr.10: 59238543-59238543 on Build GRCh38
rs755452082 Chr.10: 59238577-59238578 on Build GRCh38
rs1049174058 Chr.10: 59238439-59238439 on Build GRCh38
rs561512582 Chr.10: 59238573-59238573 on Build GRCh38
rs545551878 Chr.10: 59238494-59238494 on Build GRCh38
rs199542929 Chr.10: 59238534-59238534 on Build GRCh38
rs943319700 Chr.10: 59238479-59238480 on Build GRCh38
rs945466698 Chr.10: 59238473-59238473 on Build GRCh38
rs776627275 Chr.10: 59238620-59238620 on Build GRCh38
rs903849528 Chr.10: 59238518-59238518 on Build GRCh38
rs748240368 Chr.10: 59238594-59238594 on Build GRCh38
rs771105636 Chr.10: 59238603-59238603 on Build GRCh38
rs368184914 Chr.10: 59238548-59238548 on Build GRCh38

View Full Product Details