Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 127287451-127287722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RNF146
Entrez Gene ID
81847
Gene Name
ring finger protein 146
Gene Aliases
-
UniGene
Hs.267120
Gene Chromosome Location
Chr.6: 127266340-127289893 on Build GRCh38
Transcripts
NM_001242844.1 NM_001242845.1 NM_001242846.1 NM_001242847.1 NM_001242848.1 NM_001242849.1 NM_001242850.1 NM_001242851.1 NM_001242852.1 NM_030963.3 XM_006715571.3 XM_011536161.2 XM_011536162.2 XM_011536163.2 XM_011536164.2 XM_017011336.1 XM_017011337.1 XM_017011338.1 XM_017011339.1 XM_017011340.1 XM_017011341.1 XM_017011342.1 XM_017011343.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3858071 c.995G>A p.R332Q Chr.6: 127287611-127287611 on Build GRCh38
1072976 c.979C>T p.R327* Chr.6: 127287595-127287595 on Build GRCh38
1072975 c.970C>T p.R324* Chr.6: 127287586-127287586 on Build GRCh38
739317 c.1010G>A p.G337D Chr.6: 127287626-127287626 on Build GRCh38
7304452 c.967G>A p.D323N Chr.6: 127287583-127287583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749991765 Chr.6: 127287674-127287674 on Build GRCh38
rs775391334 Chr.6: 127287620-127287620 on Build GRCh38
rs749164556 Chr.6: 127287551-127287551 on Build GRCh38
rs111808127 Chr.6: 127287583-127287583 on Build GRCh38
rs537855606 Chr.6: 127287680-127287680 on Build GRCh38
rs776982531 Chr.6: 127287527-127287528 on Build GRCh38
rs755999333 Chr.6: 127287556-127287556 on Build GRCh38
rs143742264 Chr.6: 127287611-127287611 on Build GRCh38
rs770171920 Chr.6: 127287591-127287591 on Build GRCh38
rs764129860 Chr.6: 127287565-127287565 on Build GRCh38
rs759985928 Chr.6: 127287534-127287536 on Build GRCh38
rs773819934 Chr.6: 127287596-127287596 on Build GRCh38
rs377271416 Chr.6: 127287506-127287506 on Build GRCh38
rs371143357 Chr.6: 127287625-127287625 on Build GRCh38
rs546749357 Chr.6: 127287488-127287488 on Build GRCh38
rs779724130 Chr.6: 127287695-127287695 on Build GRCh38
rs756809979 Chr.6: 127287579-127287579 on Build GRCh38
rs771910185 Chr.6: 127287539-127287539 on Build GRCh38
rs1053669224 Chr.6: 127287600-127287600 on Build GRCh38
rs767237415 Chr.6: 127287582-127287583 on Build GRCh38
rs752982127 Chr.6: 127287697-127287697 on Build GRCh38
rs568641274 Chr.6: 127287582-127287582 on Build GRCh38
rs774652896 Chr.6: 127287610-127287610 on Build GRCh38
rs765505242 Chr.6: 127287582-127287609 on Build GRCh38
rs926454725 Chr.6: 127287590-127287590 on Build GRCh38
rs973650958 Chr.6: 127287495-127287495 on Build GRCh38
rs767427529 Chr.6: 127287543-127287543 on Build GRCh38
rs933207045 Chr.6: 127287666-127287666 on Build GRCh38
rs1009563274 Chr.6: 127287640-127287643 on Build GRCh38
rs1051623862 Chr.6: 127287677-127287677 on Build GRCh38
rs753262038 Chr.6: 127287575-127287575 on Build GRCh38
rs1012273525 Chr.6: 127287492-127287492 on Build GRCh38
rs375457836 Chr.6: 127287638-127287638 on Build GRCh38
rs760564081 Chr.6: 127287624-127287624 on Build GRCh38
rs982538097 Chr.6: 127287513-127287513 on Build GRCh38
rs778497238 Chr.6: 127287581-127287581 on Build GRCh38
rs148144282 Chr.6: 127287642-127287642 on Build GRCh38
rs748615447 Chr.6: 127287587-127287587 on Build GRCh38
rs753047902 Chr.6: 127287583-127287591 on Build GRCh38
rs756972157 Chr.6: 127287583-127287606 on Build GRCh38
rs768293915 Chr.6: 127287570-127287570 on Build GRCh38
rs552178684 Chr.6: 127287542-127287542 on Build GRCh38
rs771170597 Chr.6: 127287599-127287599 on Build GRCh38
rs764739440 Chr.6: 127287647-127287647 on Build GRCh38
rs749972364 Chr.6: 127287633-127287636 on Build GRCh38
rs145946705 Chr.6: 127287508-127287508 on Build GRCh38
rs149272268 Chr.6: 127287598-127287598 on Build GRCh38
rs779731781 Chr.6: 127287586-127287586 on Build GRCh38
rs763133290 Chr.6: 127287480-127287480 on Build GRCh38

View Full Product Details