Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 231528125-231528374 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NMUR1
Entrez Gene ID
10316
Gene Name
neuromedin U receptor 1
Gene Aliases
(FM-3), FM-3, FM3, GPC-R, GPR66, NMU1R
UniGene
Hs.471619
Gene Chromosome Location
Chr.2: 231519903-231530471 on Build GRCh38
Transcripts
NM_006056.4 XM_006712195.3 XM_006712196.3 XM_011510487.2 XM_011510488.2 XM_011510489.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
322062 c.875G>A p.R292Q Chr.2: 231528146-231528146 on Build GRCh38
1017763 c.769C>T p.R257* Chr.2: 231528252-231528252 on Build GRCh38
5797695 c.711G>T p.L237L Chr.2: 231528310-231528310 on Build GRCh38
442473 c.821C>T p.S274F Chr.2: 231528200-231528200 on Build GRCh38
6727480 c.648A>G p.S216S Chr.2: 231528373-231528373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932920452 Chr.2: 231528159-231528159 on Build GRCh38
rs749642766 Chr.2: 231528200-231528200 on Build GRCh38
rs767710968 Chr.2: 231528347-231528347 on Build GRCh38
rs745649653 Chr.2: 231528353-231528353 on Build GRCh38
rs753654709 Chr.2: 231528156-231528156 on Build GRCh38
rs754140599 Chr.2: 231528227-231528229 on Build GRCh38
rs779195143 Chr.2: 231528300-231528302 on Build GRCh38
rs373556272 Chr.2: 231528314-231528314 on Build GRCh38
rs751738088 Chr.2: 231528166-231528166 on Build GRCh38
rs147387907 Chr.2: 231528354-231528354 on Build GRCh38
rs765022352 Chr.2: 231528252-231528252 on Build GRCh38
rs750909422 Chr.2: 231528318-231528319 on Build GRCh38
rs199618212 Chr.2: 231528348-231528348 on Build GRCh38
rs749740454 Chr.2: 231528277-231528277 on Build GRCh38
rs370748156 Chr.2: 231528213-231528213 on Build GRCh38
rs746505696 Chr.2: 231528313-231528313 on Build GRCh38
rs762702531 Chr.2: 231528344-231528344 on Build GRCh38
rs780583411 Chr.2: 231528197-231528197 on Build GRCh38
rs779290553 Chr.2: 231528286-231528286 on Build GRCh38
rs745976581 Chr.2: 231528183-231528183 on Build GRCh38
rs771645983 Chr.2: 231528235-231528235 on Build GRCh38
rs774585282 Chr.2: 231528214-231528214 on Build GRCh38
rs200334481 Chr.2: 231528349-231528349 on Build GRCh38
rs928086297 Chr.2: 231528176-231528176 on Build GRCh38
rs200462040 Chr.2: 231528157-231528157 on Build GRCh38
rs756241506 Chr.2: 231528271-231528271 on Build GRCh38
rs369219078 Chr.2: 231528153-231528153 on Build GRCh38
rs757646046 Chr.2: 231528286-231528286 on Build GRCh38
rs760642418 Chr.2: 231528147-231528147 on Build GRCh38
rs767764298 Chr.2: 231528267-231528267 on Build GRCh38
rs138714852 Chr.2: 231528325-231528325 on Build GRCh38
rs770251123 Chr.2: 231528185-231528185 on Build GRCh38
rs772810450 Chr.2: 231528238-231528238 on Build GRCh38
rs748947939 Chr.2: 231528152-231528152 on Build GRCh38
rs150482580 Chr.2: 231528246-231528246 on Build GRCh38
rs766397486 Chr.2: 231528243-231528243 on Build GRCh38
rs750476073 Chr.2: 231528269-231528269 on Build GRCh38
rs766392313 Chr.2: 231528351-231528351 on Build GRCh38
rs760301353 Chr.2: 231528242-231528242 on Build GRCh38
rs371287738 Chr.2: 231528260-231528260 on Build GRCh38
rs773048181 Chr.2: 231528295-231528295 on Build GRCh38
rs769135315 Chr.2: 231528207-231528207 on Build GRCh38
rs781282455 Chr.2: 231528178-231528178 on Build GRCh38
rs748494006 Chr.2: 231528290-231528290 on Build GRCh38
rs771726239 Chr.2: 231528291-231528291 on Build GRCh38
rs759469043 Chr.2: 231528316-231528316 on Build GRCh38
rs755457086 Chr.2: 231528282-231528282 on Build GRCh38
rs764340300 Chr.2: 231528153-231528153 on Build GRCh38
rs776776103 Chr.2: 231528245-231528245 on Build GRCh38
rs757458247 Chr.2: 231528265-231528265 on Build GRCh38
rs965547383 Chr.2: 231528287-231528287 on Build GRCh38
rs147521186 Chr.2: 231528276-231528276 on Build GRCh38
rs765117659 Chr.2: 231528321-231528321 on Build GRCh38
rs367688643 Chr.2: 231528315-231528315 on Build GRCh38
rs757431516 Chr.2: 231528174-231528174 on Build GRCh38
rs747781095 Chr.2: 231528230-231528230 on Build GRCh38
rs201008110 Chr.2: 231528270-231528270 on Build GRCh38

View Full Product Details