Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 3664085-3664322 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNASE1
Entrez Gene ID
1773
Gene Name
deoxyribonuclease I
Gene Aliases
DNL1, DRNI
UniGene
Hs.629638
Gene Chromosome Location
Chr.16: 3611705-3665472 on Build GRCh38
Transcripts
XM_005255148.3 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022996.1 XM_017022997.1 XM_017022998.1 XM_017023000.1 XM_017023001.1 XM_017023002.1 XM_017023003.1 XM_017023004.1 XM_017023005.1 XM_017023006.1 NM_001272049.1 NM_016292.2 XM_011522345.1 XM_017022851.1

Gene Symbol
TRAP1
Entrez Gene ID
10131
Gene Name
TNF receptor associated protein 1
Gene Aliases
HSP 75, HSP75, HSP90L, TRAP-1
UniGene
Hs.30345
Gene Chromosome Location
Chr.16: 3658037-3717597 on Build GRCh38
Transcripts
XM_005255148.3 XM_017022992.1 XM_017022993.1 XM_017022994.1 XM_017022995.1 XM_017022996.1 XM_017022997.1 XM_017022998.1 XM_017023000.1 XM_017023001.1 XM_017023002.1 XM_017023003.1 XM_017023004.1 XM_017023005.1 XM_017023006.1 NM_001272049.1 NM_016292.2 XM_011522345.1 XM_017022851.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4656209 c.1565C>G p.T522R Chr.16: 3664278-3664278 on Build GRCh38
4434966 c.1530C>T p.H510H Chr.16: 3664313-3664313 on Build GRCh38
7394847 c.1567G>A p.E523K Chr.16: 3664276-3664276 on Build GRCh38
4060679 c.1527G>A p.E509E Chr.16: 3664316-3664316 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs556749922 Chr.16: 3664244-3664244 on Build GRCh38
rs1004761565 Chr.16: 3664134-3664134 on Build GRCh38
rs758045960 Chr.16: 3664185-3664185 on Build GRCh38
rs1043338530 Chr.16: 3664149-3664149 on Build GRCh38
rs779147132 Chr.16: 3664233-3664233 on Build GRCh38
rs761798323 Chr.16: 3664163-3664163 on Build GRCh38
rs781132432 Chr.16: 3664252-3664252 on Build GRCh38
rs114728738 Chr.16: 3664120-3664120 on Build GRCh38
rs1004979162 Chr.16: 3664187-3664187 on Build GRCh38
rs745903267 Chr.16: 3664285-3664285 on Build GRCh38
rs887335320 Chr.16: 3664273-3664273 on Build GRCh38
rs754302006 Chr.16: 3664234-3664234 on Build GRCh38
rs536877144 Chr.16: 3664236-3664236 on Build GRCh38
rs752817928 Chr.16: 3664167-3664167 on Build GRCh38
rs912954604 Chr.16: 3664276-3664276 on Build GRCh38
rs1004410095 Chr.16: 3664277-3664277 on Build GRCh38
rs934527928 Chr.16: 3664218-3664218 on Build GRCh38
rs777525310 Chr.16: 3664266-3664266 on Build GRCh38
rs772068892 Chr.16: 3664302-3664302 on Build GRCh38
rs1015396297 Chr.16: 3664191-3664191 on Build GRCh38
rs988596100 Chr.16: 3664274-3664274 on Build GRCh38
rs369558035 Chr.16: 3664262-3664262 on Build GRCh38
rs551840610 Chr.16: 3664133-3664133 on Build GRCh38
rs1019730454 Chr.16: 3664147-3664147 on Build GRCh38
rs377047464 Chr.16: 3664209-3664209 on Build GRCh38
rs532855661 Chr.16: 3664130-3664130 on Build GRCh38
rs7193381 Chr.16: 3664269-3664269 on Build GRCh38
rs115291551 Chr.16: 3664245-3664245 on Build GRCh38
rs1044079322 Chr.16: 3664251-3664251 on Build GRCh38
rs768002695 Chr.16: 3664226-3664226 on Build GRCh38
rs947019355 Chr.16: 3664148-3664148 on Build GRCh38
rs751496020 Chr.16: 3664124-3664124 on Build GRCh38
rs749142527 Chr.16: 3664270-3664270 on Build GRCh38
rs759370461 Chr.16: 3664281-3664284 on Build GRCh38
rs753989266 Chr.16: 3664177-3664177 on Build GRCh38
rs774441134 Chr.16: 3664283-3664283 on Build GRCh38
rs753159652 Chr.16: 3664229-3664229 on Build GRCh38
rs978707091 Chr.16: 3664198-3664198 on Build GRCh38
rs776028001 Chr.16: 3664303-3664303 on Build GRCh38
rs565679012 Chr.16: 3664176-3664176 on Build GRCh38
rs900743239 Chr.16: 3664152-3664152 on Build GRCh38
rs767468375 Chr.16: 3664284-3664286 on Build GRCh38
rs376835345 Chr.16: 3664263-3664263 on Build GRCh38
rs567890487 Chr.16: 3664225-3664225 on Build GRCh38
rs376389177 Chr.16: 3664114-3664115 on Build GRCh38
rs554477905 Chr.16: 3664294-3664294 on Build GRCh38
rs763883188 Chr.16: 3664119-3664119 on Build GRCh38
rs867764907 Chr.16: 3664200-3664200 on Build GRCh38
rs771142373 Chr.16: 3664282-3664282 on Build GRCh38
rs761074279 Chr.16: 3664232-3664232 on Build GRCh38
rs367688163 Chr.16: 3664264-3664264 on Build GRCh38
rs528164308 Chr.16: 3664195-3664195 on Build GRCh38
rs777797501 Chr.16: 3664227-3664227 on Build GRCh38

View Full Product Details