Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 85267785-85268058 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA13
Entrez Gene ID
377677
Gene Name
carbonic anhydrase 13
Gene Aliases
CAXIII
UniGene
Hs.127189
Gene Chromosome Location
Chr.8: 85245487-85284073 on Build GRCh38
Transcripts
NM_198584.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1635883 c.472C>T p.L158L Chr.8: 85267923-85267923 on Build GRCh38
5935376 c.491C>T p.T164I Chr.8: 85267942-85267942 on Build GRCh38
7303172 c.483T>G p.I161M Chr.8: 85267934-85267934 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs749235237 Chr.8: 85267972-85267972 on Build GRCh38
rs530670562 Chr.8: 85267849-85267849 on Build GRCh38
rs570163265 Chr.8: 85267920-85267920 on Build GRCh38
rs755327271 Chr.8: 85267878-85267878 on Build GRCh38
rs777443976 Chr.8: 85267966-85267966 on Build GRCh38
rs771749785 Chr.8: 85267858-85267858 on Build GRCh38
rs752896137 Chr.8: 85267953-85267953 on Build GRCh38
rs755231183 Chr.8: 85267926-85267926 on Build GRCh38
rs954223089 Chr.8: 85267935-85267935 on Build GRCh38
rs968531265 Chr.8: 85267959-85267959 on Build GRCh38
rs1031227629 Chr.8: 85267831-85267831 on Build GRCh38
rs763563575 Chr.8: 85267910-85267910 on Build GRCh38
rs765549891 Chr.8: 85267884-85267884 on Build GRCh38
rs781517647 Chr.8: 85267936-85267936 on Build GRCh38
rs529454463 Chr.8: 85267993-85267993 on Build GRCh38
rs1000629537 Chr.8: 85267811-85267812 on Build GRCh38
rs755872579 Chr.8: 85267964-85267964 on Build GRCh38
rs564865213 Chr.8: 85267812-85267812 on Build GRCh38
rs998360740 Chr.8: 85267818-85267818 on Build GRCh38
rs372353089 Chr.8: 85267867-85267867 on Build GRCh38
rs956325818 Chr.8: 85267868-85267868 on Build GRCh38
rs773073385 Chr.8: 85267864-85267864 on Build GRCh38
rs867901834 Chr.8: 85267929-85267929 on Build GRCh38
rs371592408 Chr.8: 85267934-85267934 on Build GRCh38
rs368124926 Chr.8: 85267919-85267919 on Build GRCh38
rs564900875 Chr.8: 85268013-85268013 on Build GRCh38
rs34047437 Chr.8: 85267907-85267907 on Build GRCh38

View Full Product Details