Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 68157774-68158015 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KMT5B
Entrez Gene ID
51111
Gene Name
lysine methyltransferase 5B
Gene Aliases
CGI-85, CGI85, SUV420H1
UniGene
Hs.632120
Gene Chromosome Location
Chr.11: 68154863-68213904 on Build GRCh38
Transcripts
NM_001300907.1 NM_001300908.1 NM_017635.4 XM_005274035.3 XM_005274036.3 XM_006718581.1 XM_011545091.1 XM_011545092.2 XM_011545093.2 XM_017017876.1 XM_017017877.1 XM_017017878.1 XM_017017879.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6519296 c.2502C>T p.G834G Chr.11: 68157844-68157844 on Build GRCh38
6519297 c.2502C>T p.G834G Chr.11: 68157844-68157844 on Build GRCh38
4908003 c.2452T>C p.Y818H Chr.11: 68157894-68157894 on Build GRCh38
4866188 c.2347C>T p.R783* Chr.11: 68157999-68157999 on Build GRCh38
5487439 c.2434C>T p.R812* Chr.11: 68157912-68157912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs878853164 Chr.11: 68157849-68157849 on Build GRCh38
rs997787256 Chr.11: 68157821-68157821 on Build GRCh38
rs377086091 Chr.11: 68157911-68157911 on Build GRCh38
rs775068563 Chr.11: 68157970-68157970 on Build GRCh38
rs374854320 Chr.11: 68157822-68157822 on Build GRCh38
rs774200646 Chr.11: 68157810-68157810 on Build GRCh38
rs774241893 Chr.11: 68157980-68157980 on Build GRCh38
rs749955049 Chr.11: 68157854-68157856 on Build GRCh38
rs745446918 Chr.11: 68157951-68157951 on Build GRCh38
rs914188764 Chr.11: 68157920-68157920 on Build GRCh38
rs775258652 Chr.11: 68157815-68157815 on Build GRCh38
rs978927620 Chr.11: 68157852-68157852 on Build GRCh38
rs757984823 Chr.11: 68157862-68157864 on Build GRCh38
rs371381032 Chr.11: 68157922-68157922 on Build GRCh38
rs764684646 Chr.11: 68157836-68157838 on Build GRCh38
rs371668215 Chr.11: 68157805-68157805 on Build GRCh38
rs149821065 Chr.11: 68157811-68157811 on Build GRCh38
rs757828667 Chr.11: 68157918-68157918 on Build GRCh38
rs765487096 Chr.11: 68157880-68157880 on Build GRCh38
rs768299849 Chr.11: 68157978-68157978 on Build GRCh38
rs755239289 Chr.11: 68157865-68157865 on Build GRCh38
rs761209606 Chr.11: 68157830-68157832 on Build GRCh38
rs377741521 Chr.11: 68157815-68157817 on Build GRCh38
rs780664772 Chr.11: 68157937-68157937 on Build GRCh38
rs139757555 Chr.11: 68157877-68157877 on Build GRCh38
rs748995814 Chr.11: 68157973-68157973 on Build GRCh38
rs774111656 Chr.11: 68157838-68157838 on Build GRCh38
rs778230550 Chr.11: 68157906-68157906 on Build GRCh38
rs766537149 Chr.11: 68157844-68157844 on Build GRCh38
rs776154444 Chr.11: 68157795-68157795 on Build GRCh38
rs758639921 Chr.11: 68157887-68157887 on Build GRCh38
rs376851966 Chr.11: 68157939-68157939 on Build GRCh38
rs145888264 Chr.11: 68157843-68157843 on Build GRCh38
rs752979595 Chr.11: 68157882-68157882 on Build GRCh38
rs769300408 Chr.11: 68157963-68157963 on Build GRCh38
rs754021856 Chr.11: 68157851-68157851 on Build GRCh38
rs763967281 Chr.11: 68157834-68157834 on Build GRCh38

View Full Product Details