Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 55663557-55663812 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
U2AF2
Entrez Gene ID
11338
Gene Name
U2 small nuclear RNA auxiliary factor 2
Gene Aliases
U2AF65
UniGene
Hs.528007
Gene Chromosome Location
Chr.19: 55654050-55674716 on Build GRCh38
Transcripts
NM_001012478.1 NM_007279.2 XM_006722994.2 XM_011526410.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4403642 c.620A>G p.E207G Chr.19: 55663622-55663622 on Build GRCh38
1681385 c.631G>A p.A211T Chr.19: 55663633-55663633 on Build GRCh38
1177425 c.702G>A p.P234P Chr.19: 55663704-55663704 on Build GRCh38
7131554 c.607C>A p.R203S Chr.19: 55663609-55663609 on Build GRCh38
1001596 c.630G>T p.Q210H Chr.19: 55663632-55663632 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs775180554 Chr.19: 55663786-55663786 on Build GRCh38
rs200495335 Chr.19: 55663752-55663752 on Build GRCh38
rs780169530 Chr.19: 55663743-55663743 on Build GRCh38
rs2303087 Chr.19: 55663781-55663781 on Build GRCh38
rs763907375 Chr.19: 55663692-55663692 on Build GRCh38
rs938556913 Chr.19: 55663617-55663617 on Build GRCh38
rs756274625 Chr.19: 55663732-55663732 on Build GRCh38
rs1027866390 Chr.19: 55663770-55663770 on Build GRCh38
rs386811093 Chr.19: 55663781-55663783 on Build GRCh38
rs372872498 Chr.19: 55663595-55663595 on Build GRCh38
rs774297269 Chr.19: 55663791-55663791 on Build GRCh38
rs774815420 Chr.19: 55663600-55663600 on Build GRCh38
rs769196337 Chr.19: 55663755-55663755 on Build GRCh38
rs310444 Chr.19: 55663783-55663783 on Build GRCh38
rs758353091 Chr.19: 55663579-55663579 on Build GRCh38
rs950844856 Chr.19: 55663776-55663777 on Build GRCh38
rs758476542 Chr.19: 55663683-55663683 on Build GRCh38
rs372237608 Chr.19: 55663704-55663704 on Build GRCh38
rs757540787 Chr.19: 55663586-55663586 on Build GRCh38
rs759196766 Chr.19: 55663641-55663641 on Build GRCh38
rs188818362 Chr.19: 55663793-55663793 on Build GRCh38
rs751454134 Chr.19: 55663792-55663792 on Build GRCh38
rs775485746 Chr.19: 55663594-55663594 on Build GRCh38
rs779307673 Chr.19: 55663760-55663760 on Build GRCh38
rs772326480 Chr.19: 55663765-55663765 on Build GRCh38
rs773566721 Chr.19: 55663771-55663771 on Build GRCh38
rs781250989 Chr.19: 55663588-55663588 on Build GRCh38
rs769895720 Chr.19: 55663593-55663593 on Build GRCh38
rs139711814 Chr.19: 55663731-55663731 on Build GRCh38
rs751420977 Chr.19: 55663697-55663697 on Build GRCh38
rs962498415 Chr.19: 55663665-55663665 on Build GRCh38
rs376049723 Chr.19: 55663620-55663620 on Build GRCh38
rs769142943 Chr.19: 55663599-55663599 on Build GRCh38
rs367695916 Chr.19: 55663589-55663589 on Build GRCh38
rs749332239 Chr.19: 55663754-55663754 on Build GRCh38
rs368338326 Chr.19: 55663674-55663674 on Build GRCh38
rs776177707 Chr.19: 55663623-55663623 on Build GRCh38
rs891005492 Chr.19: 55663602-55663602 on Build GRCh38
rs747643571 Chr.19: 55663644-55663644 on Build GRCh38
rs149231531 Chr.19: 55663608-55663608 on Build GRCh38
rs576912690 Chr.19: 55663680-55663680 on Build GRCh38
rs62126975 Chr.19: 55663730-55663730 on Build GRCh38
rs371696032 Chr.19: 55663788-55663788 on Build GRCh38
rs1018317000 Chr.19: 55663624-55663624 on Build GRCh38
rs781003609 Chr.19: 55663725-55663725 on Build GRCh38
rs902351170 Chr.19: 55663585-55663585 on Build GRCh38
rs746776887 Chr.19: 55663581-55663581 on Build GRCh38
rs777768357 Chr.19: 55663580-55663580 on Build GRCh38

View Full Product Details