Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 122262228-122262467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CASR
Entrez Gene ID
846
Gene Name
calcium sensing receptor
Gene Aliases
CAR, EIG8, FHH, FIH, GPRC2A, HHC, HHC1, HYPOC1, NSHPT, PCAR1
UniGene
Hs.435615
Gene Chromosome Location
Chr.3: 122183683-122286503 on Build GRCh38
Transcripts
NM_000388.3 NM_001178065.1 XM_005247837.2 XM_006713789.3 XM_017007324.1 XM_017007325.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1037090 c.1212C>T p.V404V Chr.3: 122262247-122262247 on Build GRCh38
1037091 c.1243C>T p.R415W Chr.3: 122262278-122262278 on Build GRCh38
7290798 c.1340G>T p.G447V Chr.3: 122262375-122262375 on Build GRCh38
1037092 c.1341C>T p.G447G Chr.3: 122262376-122262376 on Build GRCh38
1199675 c.1340G>A p.G447D Chr.3: 122262375-122262375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs895543540 Chr.3: 122262318-122262318 on Build GRCh38
rs193922421 Chr.3: 122262279-122262279 on Build GRCh38
rs766477389 Chr.3: 122262406-122262406 on Build GRCh38
rs754075314 Chr.3: 122262301-122262303 on Build GRCh38
rs1029613003 Chr.3: 122262395-122262395 on Build GRCh38
rs199511990 Chr.3: 122262332-122262332 on Build GRCh38
rs201520875 Chr.3: 122262323-122262323 on Build GRCh38
rs78925326 Chr.3: 122262414-122262414 on Build GRCh38
rs748460866 Chr.3: 122262313-122262313 on Build GRCh38
rs779350218 Chr.3: 122262420-122262420 on Build GRCh38
rs758703660 Chr.3: 122262430-122262430 on Build GRCh38
rs267599574 Chr.3: 122262362-122262362 on Build GRCh38
rs771741512 Chr.3: 122262316-122262316 on Build GRCh38
rs777348512 Chr.3: 122262436-122262436 on Build GRCh38
rs141315218 Chr.3: 122262375-122262375 on Build GRCh38
rs765170020 Chr.3: 122262352-122262352 on Build GRCh38
rs762799076 Chr.3: 122262373-122262373 on Build GRCh38
rs142818334 Chr.3: 122262320-122262320 on Build GRCh38
rs756206452 Chr.3: 122262389-122262389 on Build GRCh38
rs12493789 Chr.3: 122262368-122262368 on Build GRCh38
rs540006567 Chr.3: 122262386-122262386 on Build GRCh38
rs375297380 Chr.3: 122262412-122262412 on Build GRCh38
rs200443784 Chr.3: 122262271-122262271 on Build GRCh38
rs200243816 Chr.3: 122262355-122262355 on Build GRCh38
rs200650843 Chr.3: 122262429-122262429 on Build GRCh38
rs977386230 Chr.3: 122262254-122262254 on Build GRCh38
rs199824580 Chr.3: 122262415-122262415 on Build GRCh38
rs746515147 Chr.3: 122262322-122262322 on Build GRCh38

View Full Product Details