Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 41432195-41432448 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP5SL
Entrez Gene ID
55101
Gene Name
ATP5S like
Gene Aliases
-
UniGene
Hs.351099
Gene Chromosome Location
Chr.19: 41431318-41447822 on Build GRCh38
Transcripts
NM_001167867.1 NM_001167868.1 NM_001167869.1 NM_001167870.1 NM_001167871.1 NM_001320838.1 NM_001320840.1 NM_001320841.1 NM_018035.2 NR_030765.1 NR_135476.1 NR_135477.1 NR_135478.1 XM_011527065.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4636365 c.745C>T p.R249W Chr.19: 41432260-41432260 on Build GRCh38
7130126 c.624C>A p.D208E Chr.19: 41432381-41432381 on Build GRCh38
5464595 c.600C>T p.N200N Chr.19: 41432405-41432405 on Build GRCh38
6448382 c.672G>A p.E224E Chr.19: 41432333-41432333 on Build GRCh38
4505632 c.680C>T p.P227L Chr.19: 41432325-41432325 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782571130 Chr.19: 41432382-41432382 on Build GRCh38
rs782152400 Chr.19: 41432424-41432424 on Build GRCh38
rs782375189 Chr.19: 41432417-41432417 on Build GRCh38
rs571205426 Chr.19: 41432315-41432315 on Build GRCh38
rs782050291 Chr.19: 41432262-41432262 on Build GRCh38
rs782524437 Chr.19: 41432307-41432307 on Build GRCh38
rs782558606 Chr.19: 41432379-41432379 on Build GRCh38
rs782728447 Chr.19: 41432269-41432269 on Build GRCh38
rs782440252 Chr.19: 41432302-41432302 on Build GRCh38
rs782160432 Chr.19: 41432296-41432296 on Build GRCh38
rs1040904774 Chr.19: 41432276-41432276 on Build GRCh38
rs781957938 Chr.19: 41432270-41432270 on Build GRCh38
rs534167049 Chr.19: 41432372-41432372 on Build GRCh38
rs782387270 Chr.19: 41432395-41432395 on Build GRCh38
rs782720309 Chr.19: 41432364-41432364 on Build GRCh38
rs185136417 Chr.19: 41432385-41432385 on Build GRCh38
rs201728769 Chr.19: 41432322-41432322 on Build GRCh38
rs143580973 Chr.19: 41432367-41432367 on Build GRCh38
rs200395698 Chr.19: 41432215-41432215 on Build GRCh38
rs143331885 Chr.19: 41432384-41432384 on Build GRCh38
rs782665880 Chr.19: 41432400-41432400 on Build GRCh38
rs781992030 Chr.19: 41432256-41432256 on Build GRCh38
rs782798701 Chr.19: 41432289-41432289 on Build GRCh38
rs199654508 Chr.19: 41432260-41432260 on Build GRCh38
rs782191910 Chr.19: 41432252-41432252 on Build GRCh38
rs782544246 Chr.19: 41432285-41432285 on Build GRCh38
rs1002961129 Chr.19: 41432326-41432326 on Build GRCh38
rs538133275 Chr.19: 41432274-41432274 on Build GRCh38
rs781933529 Chr.19: 41432406-41432406 on Build GRCh38
rs2231943 Chr.19: 41432317-41432317 on Build GRCh38
rs782269362 Chr.19: 41432223-41432223 on Build GRCh38
rs782212578 Chr.19: 41432321-41432321 on Build GRCh38
rs1050704343 Chr.19: 41432312-41432312 on Build GRCh38
rs201804717 Chr.19: 41432318-41432318 on Build GRCh38
rs138402902 Chr.19: 41432352-41432352 on Build GRCh38
rs782366585 Chr.19: 41432255-41432255 on Build GRCh38
rs781867955 Chr.19: 41432277-41432277 on Build GRCh38
rs782041670 Chr.19: 41432362-41432362 on Build GRCh38
rs143413450 Chr.19: 41432239-41432239 on Build GRCh38
rs782238272 Chr.19: 41432391-41432391 on Build GRCh38
rs147192262 Chr.19: 41432378-41432378 on Build GRCh38
rs369998203 Chr.19: 41432293-41432293 on Build GRCh38
rs782130501 Chr.19: 41432328-41432328 on Build GRCh38
rs1043444442 Chr.19: 41432222-41432222 on Build GRCh38
rs2231944 Chr.19: 41432303-41432303 on Build GRCh38
rs1035230097 Chr.19: 41432333-41432333 on Build GRCh38
rs371036948 Chr.19: 41432369-41432369 on Build GRCh38
rs962413661 Chr.19: 41432323-41432323 on Build GRCh38
rs782554050 Chr.19: 41432242-41432242 on Build GRCh38
rs377558416 Chr.19: 41432356-41432356 on Build GRCh38
rs782076798 Chr.19: 41432409-41432409 on Build GRCh38
rs201587564 Chr.19: 41432401-41432401 on Build GRCh38
rs782006787 Chr.19: 41432421-41432421 on Build GRCh38
rs148293886 Chr.19: 41432273-41432273 on Build GRCh38
rs782673118 Chr.19: 41432311-41432311 on Build GRCh38
rs149741041 Chr.19: 41432390-41432390 on Build GRCh38
rs782307590 Chr.19: 41432404-41432404 on Build GRCh38

View Full Product Details