Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 168063650-168063923 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD1
Entrez Gene ID
79981
Gene Name
FERM domain containing 1
Gene Aliases
bA164L23.1
UniGene
Hs.266746
Gene Chromosome Location
Chr.6: 168055745-168093007 on Build GRCh38
Transcripts
NM_001122841.2 NM_024919.4 NR_110312.1 XM_011536137.1 XM_011536138.1 XM_011536140.1 XM_011536141.1 XM_011536142.2 XM_011536143.1 XM_011536145.1 XM_011536146.2 XM_017011317.1 XM_017011318.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5599577 c.678C>T p.I226I Chr.6: 168063727-168063727 on Build GRCh38
4503938 c.651C>T p.I217I Chr.6: 168063754-168063754 on Build GRCh38
261008 c.718C>A p.L240M Chr.6: 168063687-168063687 on Build GRCh38
338798 c.747C>T p.F249F Chr.6: 168063658-168063658 on Build GRCh38
741119 c.730G>A p.E244K Chr.6: 168063675-168063675 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749011762 Chr.6: 168063688-168063688 on Build GRCh38
rs779195159 Chr.6: 168063695-168063695 on Build GRCh38
rs1029993981 Chr.6: 168063815-168063815 on Build GRCh38
rs562906805 Chr.6: 168063783-168063783 on Build GRCh38
rs777996607 Chr.6: 168063768-168063768 on Build GRCh38
rs779747110 Chr.6: 168063730-168063730 on Build GRCh38
rs900399433 Chr.6: 168063792-168063792 on Build GRCh38
rs372228355 Chr.6: 168063770-168063770 on Build GRCh38
rs771406095 Chr.6: 168063771-168063771 on Build GRCh38
rs779694194 Chr.6: 168063683-168063683 on Build GRCh38
rs748495187 Chr.6: 168063696-168063696 on Build GRCh38
rs776718150 Chr.6: 168063773-168063773 on Build GRCh38
rs748463474 Chr.6: 168063745-168063745 on Build GRCh38
rs200870764 Chr.6: 168063698-168063698 on Build GRCh38
rs772486751 Chr.6: 168063697-168063697 on Build GRCh38
rs201393269 Chr.6: 168063701-168063701 on Build GRCh38
rs763158276 Chr.6: 168063742-168063742 on Build GRCh38
rs370281501 Chr.6: 168063736-168063736 on Build GRCh38
rs750430628 Chr.6: 168063729-168063729 on Build GRCh38
rs769915656 Chr.6: 168063788-168063788 on Build GRCh38
rs1014657993 Chr.6: 168063887-168063887 on Build GRCh38
rs368931831 Chr.6: 168063761-168063761 on Build GRCh38
rs768388623 Chr.6: 168063694-168063694 on Build GRCh38
rs73028318 Chr.6: 168063722-168063722 on Build GRCh38
rs78853029 Chr.6: 168063733-168063733 on Build GRCh38
rs1004636351 Chr.6: 168063829-168063829 on Build GRCh38
rs765399271 Chr.6: 168063721-168063721 on Build GRCh38
rs373702680 Chr.6: 168063743-168063743 on Build GRCh38
rs867881330 Chr.6: 168063799-168063799 on Build GRCh38
rs767371388 Chr.6: 168063723-168063723 on Build GRCh38
rs776662658 Chr.6: 168063710-168063710 on Build GRCh38
rs759576374 Chr.6: 168063787-168063787 on Build GRCh38
rs574911340 Chr.6: 168063809-168063809 on Build GRCh38
rs114522649 Chr.6: 168063828-168063828 on Build GRCh38
rs142892895 Chr.6: 168063702-168063702 on Build GRCh38
rs553570166 Chr.6: 168063719-168063719 on Build GRCh38

View Full Product Details