Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 44851189-44851456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFTUD2
Entrez Gene ID
9343
Gene Name
elongation factor Tu GTP binding domain containing 2
Gene Aliases
MFDGA, MFDM, SNRNP116, Snrp116, Snu114, U5-116KD
UniGene
Hs.151787
Gene Chromosome Location
Chr.17: 44850287-44899625 on Build GRCh38
Transcripts
NM_001142605.1 NM_001258353.1 NM_001258354.1 NM_004247.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
291557 c.2843G>T p.S948I Chr.17: 44851350-44851350 on Build GRCh38
6400264 c.2874G>T p.L958F Chr.17: 44851319-44851319 on Build GRCh38
3819753 c.2882T>G p.L961R Chr.17: 44851311-44851311 on Build GRCh38
4840556 c.2859C>T p.F953F Chr.17: 44851334-44851334 on Build GRCh38
5809624 c.2871G>T p.M957I Chr.17: 44851322-44851322 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993901302 Chr.17: 44851435-44851435 on Build GRCh38
rs760718291 Chr.17: 44851293-44851293 on Build GRCh38
rs762283826 Chr.17: 44851277-44851277 on Build GRCh38
rs575773370 Chr.17: 44851212-44851212 on Build GRCh38
rs934984150 Chr.17: 44851408-44851408 on Build GRCh38
rs528607188 Chr.17: 44851334-44851334 on Build GRCh38
rs772628768 Chr.17: 44851283-44851283 on Build GRCh38
rs374007980 Chr.17: 44851270-44851270 on Build GRCh38
rs180802836 Chr.17: 44851223-44851223 on Build GRCh38
rs759880893 Chr.17: 44851269-44851269 on Build GRCh38
rs777487912 Chr.17: 44851239-44851239 on Build GRCh38
rs748693215 Chr.17: 44851228-44851228 on Build GRCh38
rs756853277 Chr.17: 44851390-44851390 on Build GRCh38
rs748509447 Chr.17: 44851406-44851406 on Build GRCh38
rs758301115 Chr.17: 44851229-44851229 on Build GRCh38
rs868129814 Chr.17: 44851251-44851251 on Build GRCh38
rs139206848 Chr.17: 44851287-44851287 on Build GRCh38
rs375962052 Chr.17: 44851259-44851259 on Build GRCh38
rs764957956 Chr.17: 44851349-44851349 on Build GRCh38
rs772615359 Chr.17: 44851407-44851407 on Build GRCh38
rs770747714 Chr.17: 44851262-44851262 on Build GRCh38
rs898027002 Chr.17: 44851433-44851433 on Build GRCh38
rs377045327 Chr.17: 44851391-44851391 on Build GRCh38
rs752526926 Chr.17: 44851350-44851350 on Build GRCh38
rs755775517 Chr.17: 44851401-44851401 on Build GRCh38
rs777823115 Chr.17: 44851373-44851373 on Build GRCh38
rs774717614 Chr.17: 44851273-44851273 on Build GRCh38
rs369893390 Chr.17: 44851266-44851266 on Build GRCh38
rs745518184 Chr.17: 44851400-44851400 on Build GRCh38
rs770244136 Chr.17: 44851319-44851319 on Build GRCh38
rs776332971 Chr.17: 44851241-44851241 on Build GRCh38
rs773728885 Chr.17: 44851412-44851412 on Build GRCh38
rs751166725 Chr.17: 44851383-44851383 on Build GRCh38
rs778634820 Chr.17: 44851434-44851434 on Build GRCh38
rs770012393 Chr.17: 44851372-44851373 on Build GRCh38
rs902210932 Chr.17: 44851402-44851402 on Build GRCh38
rs766394225 Chr.17: 44851295-44851295 on Build GRCh38
rs1019138899 Chr.17: 44851410-44851410 on Build GRCh38
rs202167305 Chr.17: 44851331-44851331 on Build GRCh38
rs779207813 Chr.17: 44851405-44851405 on Build GRCh38
rs758339264 Chr.17: 44851372-44851372 on Build GRCh38

View Full Product Details