Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 98648691-98648918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APAF1
Entrez Gene ID
317
Gene Name
apoptotic peptidase activating factor 1
Gene Aliases
APAF-1, CED4
UniGene
Hs.552567
Gene Chromosome Location
Chr.12: 98645141-98735433 on Build GRCh38
Transcripts
NM_001160.2 NM_013229.2 NM_181861.1 NM_181868.1 NM_181869.1 XM_017019250.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4822807 c.287C>T p.S96F Chr.12: 98648774-98648774 on Build GRCh38
2047214 c.239A>T p.Y80F Chr.12: 98648726-98648726 on Build GRCh38
5937528 c.218A>T p.N73I Chr.12: 98648705-98648705 on Build GRCh38
432038 c.247C>T p.L83F Chr.12: 98648734-98648734 on Build GRCh38
5027658 c.284C>G p.S95C Chr.12: 98648771-98648771 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs745628140 Chr.12: 98648869-98648869 on Build GRCh38
rs765701526 Chr.12: 98648819-98648819 on Build GRCh38
rs139072016 Chr.12: 98648866-98648866 on Build GRCh38
rs746604749 Chr.12: 98648881-98648881 on Build GRCh38
rs771813021 Chr.12: 98648875-98648875 on Build GRCh38
rs775321405 Chr.12: 98648855-98648856 on Build GRCh38
rs755133056 Chr.12: 98648840-98648840 on Build GRCh38
rs150823984 Chr.12: 98648774-98648774 on Build GRCh38
rs925746508 Chr.12: 98648749-98648749 on Build GRCh38
rs753876879 Chr.12: 98648844-98648844 on Build GRCh38
rs528877600 Chr.12: 98648727-98648727 on Build GRCh38
rs775021247 Chr.12: 98648769-98648769 on Build GRCh38
rs768215449 Chr.12: 98648886-98648886 on Build GRCh38
rs777800292 Chr.12: 98648731-98648731 on Build GRCh38
rs978576647 Chr.12: 98648743-98648743 on Build GRCh38
rs141352935 Chr.12: 98648761-98648761 on Build GRCh38
rs917012585 Chr.12: 98648806-98648806 on Build GRCh38
rs781078640 Chr.12: 98648842-98648842 on Build GRCh38
rs770761439 Chr.12: 98648757-98648757 on Build GRCh38
rs369766511 Chr.12: 98648849-98648849 on Build GRCh38
rs565949071 Chr.12: 98648838-98648838 on Build GRCh38
rs146324917 Chr.12: 98648811-98648811 on Build GRCh38
rs941494231 Chr.12: 98648779-98648779 on Build GRCh38
rs993896526 Chr.12: 98648813-98648813 on Build GRCh38
rs768104025 Chr.12: 98648810-98648810 on Build GRCh38
rs763317313 Chr.12: 98648834-98648834 on Build GRCh38
rs779703679 Chr.12: 98648880-98648880 on Build GRCh38
rs751796312 Chr.12: 98648836-98648836 on Build GRCh38
rs749134752 Chr.12: 98648737-98648737 on Build GRCh38
rs756164693 Chr.12: 98648728-98648728 on Build GRCh38
rs139951279 Chr.12: 98648758-98648758 on Build GRCh38
rs776002033 Chr.12: 98648896-98648896 on Build GRCh38
rs750740410 Chr.12: 98648833-98648833 on Build GRCh38

View Full Product Details