Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 409267-409516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
-
UniGene
Hs.631656
Gene Chromosome Location
Chr.12: 389350-442642 on Build GRCh38
Transcripts
NM_001130146.1 NM_001130147.1 NM_001130148.1 NM_032358.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5841209 c.11C>T p.T4I Chr.12: 409394-409394 on Build GRCh38
6863906 c.17delC p.T6fs*72 Chr.12: 409400-409400 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369317610 Chr.12: 409365-409365 on Build GRCh38
rs781605518 Chr.12: 409382-409382 on Build GRCh38
rs765257911 Chr.12: 409362-409362 on Build GRCh38
rs766291102 Chr.12: 409473-409473 on Build GRCh38
rs749383721 Chr.12: 409396-409396 on Build GRCh38
rs760254593 Chr.12: 409450-409450 on Build GRCh38
rs776069385 Chr.12: 409458-409458 on Build GRCh38
rs766110274 Chr.12: 409384-409384 on Build GRCh38
rs761371661 Chr.12: 409459-409459 on Build GRCh38
rs1008710835 Chr.12: 409472-409472 on Build GRCh38
rs775095121 Chr.12: 409339-409339 on Build GRCh38
rs111544508 Chr.12: 409381-409381 on Build GRCh38
rs760305018 Chr.12: 409335-409335 on Build GRCh38
rs767210377 Chr.12: 409338-409338 on Build GRCh38
rs746275717 Chr.12: 409424-409424 on Build GRCh38
rs376809903 Chr.12: 409355-409355 on Build GRCh38
rs771402642 Chr.12: 409399-409399 on Build GRCh38
rs145608806 Chr.12: 409421-409421 on Build GRCh38
rs752997950 Chr.12: 409383-409383 on Build GRCh38
rs763531689 Chr.12: 409346-409346 on Build GRCh38
rs373007056 Chr.12: 409465-409465 on Build GRCh38
rs772460484 Chr.12: 409434-409434 on Build GRCh38
rs761658285 Chr.12: 409359-409359 on Build GRCh38
rs750299452 Chr.12: 409363-409363 on Build GRCh38
rs868716799 Chr.12: 409410-409410 on Build GRCh38
rs762942655 Chr.12: 409468-409468 on Build GRCh38
rs775780238 Chr.12: 409440-409440 on Build GRCh38
rs759885037 Chr.12: 409337-409337 on Build GRCh38
rs772772088 Chr.12: 409331-409331 on Build GRCh38
rs1037148780 Chr.12: 409428-409428 on Build GRCh38
rs768285131 Chr.12: 409457-409457 on Build GRCh38
rs199703250 Chr.12: 409342-409342 on Build GRCh38
rs758242970 Chr.12: 409368-409368 on Build GRCh38
rs558563379 Chr.12: 409410-409410 on Build GRCh38

View Full Product Details