Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 38689877-38690130 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FREM2
Entrez Gene ID
341640
Gene Name
FRAS1 related extracellular matrix protein 2
Gene Aliases
-
UniGene
Hs.253994
Gene Chromosome Location
Chr.13: 38687036-38887131 on Build GRCh38
Transcripts
NM_207361.5 XM_017020554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4617458 c.2650C>A p.L884M Chr.13: 38689994-38689994 on Build GRCh38
4821983 c.2551G>C p.E851Q Chr.13: 38689895-38689895 on Build GRCh38
4168957 c.2631G>C p.M877I Chr.13: 38689975-38689975 on Build GRCh38
4047515 c.2707G>T p.A903S Chr.13: 38690051-38690051 on Build GRCh38
5496598 c.2661G>T p.G887G Chr.13: 38690005-38690005 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs916795794 Chr.13: 38690090-38690090 on Build GRCh38
rs140353326 Chr.13: 38690098-38690098 on Build GRCh38
rs373603200 Chr.13: 38690062-38690062 on Build GRCh38
rs955235814 Chr.13: 38690041-38690041 on Build GRCh38
rs745927646 Chr.13: 38690104-38690104 on Build GRCh38
rs536688833 Chr.13: 38690093-38690093 on Build GRCh38
rs1052425955 Chr.13: 38689908-38689908 on Build GRCh38
rs754766389 Chr.13: 38690080-38690080 on Build GRCh38
rs749932417 Chr.13: 38690054-38690054 on Build GRCh38
rs764607184 Chr.13: 38690033-38690033 on Build GRCh38
rs7329939 Chr.13: 38689946-38689946 on Build GRCh38
rs753600025 Chr.13: 38690072-38690072 on Build GRCh38
rs747120941 Chr.13: 38689971-38689971 on Build GRCh38
rs755779564 Chr.13: 38689958-38689958 on Build GRCh38
rs979599972 Chr.13: 38690107-38690107 on Build GRCh38
rs113698508 Chr.13: 38690016-38690016 on Build GRCh38
rs778948730 Chr.13: 38689969-38689969 on Build GRCh38
rs775756671 Chr.13: 38690007-38690007 on Build GRCh38
rs146685625 Chr.13: 38690084-38690084 on Build GRCh38
rs781485282 Chr.13: 38690100-38690100 on Build GRCh38
rs750020230 Chr.13: 38689952-38689952 on Build GRCh38
rs763353286 Chr.13: 38690021-38690021 on Build GRCh38
rs779617548 Chr.13: 38689903-38689903 on Build GRCh38
rs767073356 Chr.13: 38690053-38690053 on Build GRCh38
rs759732621 Chr.13: 38690001-38690001 on Build GRCh38
rs752022191 Chr.13: 38690034-38690034 on Build GRCh38
rs994268140 Chr.13: 38690047-38690047 on Build GRCh38
rs61995710 Chr.13: 38689944-38689944 on Build GRCh38
rs778743216 Chr.13: 38689973-38689973 on Build GRCh38
rs149850058 Chr.13: 38689913-38689913 on Build GRCh38
rs768528556 Chr.13: 38689965-38689965 on Build GRCh38
rs770048601 Chr.13: 38690004-38690004 on Build GRCh38
rs1025301678 Chr.13: 38690055-38690055 on Build GRCh38
rs777003453 Chr.13: 38689988-38689988 on Build GRCh38
rs536960127 Chr.13: 38689904-38689904 on Build GRCh38
rs779781015 Chr.13: 38689963-38689963 on Build GRCh38
rs371536494 Chr.13: 38690060-38690060 on Build GRCh38
rs761326295 Chr.13: 38690040-38690040 on Build GRCh38
rs115142317 Chr.13: 38689909-38689909 on Build GRCh38
rs773434156 Chr.13: 38690017-38690018 on Build GRCh38
rs566743573 Chr.13: 38689940-38689940 on Build GRCh38
rs891733811 Chr.13: 38689912-38689912 on Build GRCh38
rs998092516 Chr.13: 38689937-38689937 on Build GRCh38
rs114555425 Chr.13: 38689964-38689964 on Build GRCh38

View Full Product Details